Skip to Content
Merck
All Photos(3)

Key Documents

704105

Sigma-Aldrich

Polycaprolactone

average Mn 45,000

Synonym(s):

2-Oxepanone homopolymer, 6-Caprolactone polymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O2)n
UNSPSC Code:
12162002
NACRES:
NA.21
Pricing and availability is not currently available.

form

pellets

Quality Level

melt index

1.8 g/10 min (80 °C, 44 psi)

mol wt

Mn 40,000-50,000
Mw 48,000-90,000
average Mn 45,000

mp

56-64 °C
60 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

General description

Polycaprolactone (PCL) is a semi-crystalline linear resorbable aliphatic polyester that is composed of repeating units of hexanoate. It has good mechanical properties, miscibility, and biodegradability. PCL is a soft and hard-tissue compatible material that can be used in sutures and drug delivery systems.[1][2][3]

Application

PCL can form a nanocomposite with graphene for the fabrication of biodegradable systems for bone tissue engineering.[4][5]

Features and Benefits

Biodegradable polymer
Non-toxic, biodegradable in soil, broad miscibility, mechanical compatibility with many polymers and good adhesion to a broad spectrum of substrates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Covalently linked biocompatible graphenepolycaprolactone composites for tissue engineering
Sayyar S, et al.
Carbon, 52(5), 296-304 (2013)
A novel degradable polycaprolactone networks for tissue engineering
Kweon H, et al.
Biomaterials, 24(5), 801-808 (2003)
Properties of thermoplastic blends: starch-polycaprolactone
Averous L, et al.
Polymer, 41(11), 4157-4167 (2000)
Synthesis of polycaprolactone: a review
Labet M and Thielemans W
Chemical Society Reviews, 38(12), 3484-3504 (2009)
Bone tissue engineering using polycaprolactone scaffolds fabricated via selective laser sintering
Williams JM, et al.
Biomaterials, 26(23), 4817-4827 (2005)

Articles

In the past two decades, tissue engineering and regenerative medicine have become important interdisciplinary fields that span biology, chemistry, engineering, and medicine.

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service