Skip to Content
Merck
All Photos(2)

Key Documents

920436

Sigma-Aldrich

NanoFabTx PEG-PLA drug formulation screening kit

for synthesis of PEGylated nanoparticles

Synonym(s):

Microfluidics, NanoFabTx reagent kit, Nanoformulation, Nanoparticle kit, PEG-PLA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.23
Pricing and availability is not currently available.

description

Kit components :
PEGPLA-L-M (919942-500mg)
PEGPLA-H-M (919934-500mg)
Stabilizer-Nano (907766-5g)

PEG-PLA drug formulation screening kit, for synthesis of PEGylated PLA nanoparticles

Quality Level

application(s)

advanced drug delivery

storage temp.

2-8°C

General description

NanoFabTx PEG-PLA drug formulation screening kit is a nanoformulation kit containing rationally selected PEGylated PLA polymers that have been used widely for controlled drug release of many different types of therapeutic molecules. The kit contains protocols for two different particle synthesis methods:
  • A Nanoprecipitation protocol to prepare drug-encapsulated nanoparticles in standard laboratory glassware.
  • A Microfluidics protocol using commercial platforms or syringe pumps.
The microfluidics protocol uses NanoFabTx device kits (911593), which provide all the microfluidics chips, fittings, and tubing required to get started with microfluidics-based synthesis (compatible microfluidics system or syringe pump required).

Application

NanoFabTx PEG-PLA drug formulation screening kit enables users to encapsulate a wide variety of therapeutic drug molecules in specifically sized, biodegradable, PEGylated PLA nanoparticles. By simplifying the chemistry and providing an easy-to-use toolkit with step-by-step instructions, our NanoFabTx nanoformulation screening kit helps researchers achieve reproducible synthesis of drug encapsulating nanoparticles with narrow size dispersity, eliminating the need for lengthy trial-and-error optimization. Suitable for biomedical research applications such as oncology, immuno-oncology, gene delivery, and vaccine delivery, PEG-PLA nanoparticles, made using the NanoFabTx PEG-PLA drug formulation screening kit, are designed for maximizing the encapsulation of hydrophobic drugs to minimize the time and expensive wasted reagents of synthetic optimization.

Features and Benefits

  • Includes tested protocols with step-by-step instructions for nanoprecipitation or microfluidics-based syntheses
  • Requires minimal laboratory setup
  • Optimized to achieve monodisperse, homogenously shaped, biodegradable, PEGylated PLA nanoparticles in any size between 60 and 100 nm
  • Formulated to maximize the encapsulation of hydrophobic drugs
  • Includes two different PEGylated PLAs

Legal Information

NANOFABTX is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sungmin Nam et al.
Biomaterials, 200, 15-24 (2019-02-12)
Hydrogels are commonly used as artificial extracellular matrices for 3D cell culture and for tissue engineering. Viscoelastic hydrogels with tunable stress relaxation have recently been developed, and stress relaxation in the hydrogels has been found to play a key role
S Freiberg et al.
International journal of pharmaceutics, 282(1-2), 1-18 (2004-09-01)
Polymer microspheres can be employed to deliver medication in a rate-controlled and sometimes targeted manner. Medication is released from a microsphere by drug leaching from the polymer or by degradation of the polymer matrix. Since the rate of drug release
Verónica Lassalle et al.
Macromolecular bioscience, 7(6), 767-783 (2007-06-02)
The controlled release of medicaments remains the most convenient way of drug delivery. Therefore, a wide variety of reports can be found in the open literature dealing with drug delivery systems. In particular, the use of nano- and microparticles devices
Byung Kook Lee et al.
Advanced drug delivery reviews, 107, 176-191 (2016-06-06)
Poly(d,l-lactic acid) (PLA) has been widely used for various biomedical applications for its biodegradable, biocompatible, and nontoxic properties. Various methods, such as emulsion, salting out, and precipitation, have been used to make better PLA micro- and nano-particle formulations. They are
Nazila Kamaly et al.
Chemical Society reviews, 41(7), 2971-3010 (2012-03-06)
Polymeric materials have been used in a range of pharmaceutical and biotechnology products for more than 40 years. These materials have evolved from their earlier use as biodegradable products such as resorbable sutures, orthopaedic implants, macroscale and microscale drug delivery

Articles

NanoFabTx™ platform accelerates drug development with ready-to-use formulations and microfluidic devices for particle synthesis.

NanoFabTx™ platform accelerates drug development with ready-to-use formulations and microfluidic devices for particle synthesis.

NanoFabTx™ platform accelerates drug development with ready-to-use formulations and microfluidic devices for particle synthesis.

NanoFabTx™ platform accelerates drug development with ready-to-use formulations and microfluidic devices for particle synthesis.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service