225681
4-(Diethylamino)salicylaldehyde
98%
Synonym(s):
4-Diethylamino-2-hydroxybenzaldehyde
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
(C2H5)2NC6H3-2-(OH)CHO
CAS Number:
Molecular Weight:
193.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
mp
60-62 °C (lit.)
functional group
aldehyde
amine
SMILES string
CCN(CC)c1ccc(C=O)c(O)c1
InChI
1S/C11H15NO2/c1-3-12(4-2)10-6-5-9(8-13)11(14)7-10/h5-8,14H,3-4H2,1-2H3
InChI key
XFVZSRRZZNLWBW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
4-(Diethylamino)salicylaldehyde was used in the synthesis of Schiff-base ligand by monocondensation with diaminomaleonitrile.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jean Pierre Costes et al.
Inorganic chemistry, 44(6), 1973-1982 (2005-03-15)
An H2L Schiff-base ligand that was obtained from the monocondensation of diaminomaleonitrile and 4-(diethylamino)salicylaldehyde is reported together with four related nickel(II) complexes formulated as [Ni(L)(L')] (L' = MePhCHNH2, iPrNH2, Py, and PPh3). Crystal structures have been solved for H2L, [Ni(L)(MePhCHNH2)]
Manojkumar Jadhao et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 22(1), 47-59 (2016-11-09)
Amyloid-β peptides and their metal-associated aggregated states have been implicated in the pathogenesis of Alzheimer's disease. The present paper epitomises the design and synthesis of a small, neutral, lipophilic benzothiazole Schiff base (E)-2-((6-chlorobenzo[d]thiazol-2-ylimino)methyl)-5-diethylamino)phenol (CBMDP), and explores its multifunctionalty as a
Takahiro Nomoto et al.
Science advances, 6(4), eaaz1722-eaaz1722 (2020-02-06)
In the current clinical boron neutron capture therapy (BNCT), p-boronophenylalanine (BPA) has been the most powerful drug owing to its ability to accumulate selectively within cancers through cancer-related amino acid transporters including LAT1. However, the therapeutic success of BPA has
Huirong Yao et al.
Journal of fluorescence, 25(6), 1637-1643 (2015-09-19)
A novel homodimer of the styryldehydropyridocolinium dye (TPTP) has been synthesized and characterized. Free TPTP exhibited low fluorescence quantum yield and large Stokes shift (over 160 nm) in water. However, it showed a significant fluorescence turn-on effect upon intercalation into DNA
Anti-Hypochlorite and Catalytic Activity of Commercially Available Moringa oleifera Diet Supplement.
Karolina Starzak et al.
Molecules (Basel, Switzerland), 24(18) (2019-09-25)
Aiming at the assessment of the pro-health, and especially anti-hypochlorite properties of Moringa oleifera species a representative, commercially available Moringa oleifera dietary supplement was used as a substrate for the preparation of aqueous Moringa extract. The anti-hypochlorite activity of the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service