Skip to Content
Merck
All Photos(3)

Key Documents

431788

Sigma-Aldrich

Ethylenediaminetetraacetic acid

99.995% trace metals basis

Synonym(s):

(Ethylenedinitrilo)tetraacetic acid, EDTA, Edathamil, Ethylenedinitrilotetraacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO2CCH2)2NCH2CH2N(CH2CO2H)2
CAS Number:
Molecular Weight:
292.24
Beilstein:
1716295
EC Number:
MDL number:
UNSPSC Code:
12352116
eCl@ss:
39030907
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99.995% trace metals basis

form

powder

technique(s)

electrophoresis: suitable

mp

250 °C (dec.) (lit.)

solubility

water: soluble 0.4 g/L at 20 °C

density

1.46 g/cm3 at 20 °C

functional group

amine
carboxylic acid

SMILES string

OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)

InChI key

KCXVZYZYPLLWCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylenediaminetetraacetic acid is a water-soluble chelating agent that can form complexes with metal ions.It is useful for metal ion detection and water treatment

Application

Ethylenediaminetetraacetic acid can be used:
  • As a chelating agent in the preparation of ferromagnetic BiFeO3 nanoparticles by sol-gel process.
  • As a mobile phase in chromatographic separation of rare earth elements.
  • To functionalize the Au nanoparticles, which are used as probes in the colorimetric estimation of chromium(III) ions in solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethylenediaminetetraacetic acid in endodontics
Z Mohammadi, et al.
European journal of dentistry, 7, S135-S142 (2013)
Nuclear magnetic resonance studies of metal-ethylenediaminetetraacetic acid complexes.
Kula RJ
Journal of the American Chemical Society, 85(19), 2930-2936 (1963)
Properties and Infrared Spectra of Ethylenediaminetetraacetic Acid Complexes. II. Chelates of Divalent Ions
Sawyer DT
Journal of the American Chemical Society, 81(4), 816-820 (1959)
EDTA: the chelating agent under environmental scrutiny
C Oviedo, et al.
Quimica nova, 26, 901-905 (2003)
Ethylenediaminetetraacetic acid functionalized gold nanoparticles for sensitive colorimetric detection of chromium (III)
Li J, et al.
J. Chin. Chem. Soc., 61(12), 1395-1399 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service