Select a Size
About This Item
product line
BioReagent
Quality Segment
assay
98.5-102.0%
form
powder
technique(s)
cell culture | mammalian: suitable
solubility
H2O: 160 mg/mL
shipped in
ambient
storage temp.
room temp
SMILES string
O.O.[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
InChI
1S/C10H16N2O8.4Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;;2*1H2/q;4*+1;;/p-4
InChI key
KSYNLCYTMRMCGG-UHFFFAOYSA-J
General description
Ethylenediaminetetraacetic acid (EDTA) is a chelator of metal ions. It is a substituted diamine, which has antibacterial activity. EDTA removes the undesirable effects of ferric, cupric and manganic ions in bleaching. It prevents cellular division, chlorophyll synthesis and algal biomass production. EDTA is an inhibitor of metalloprotease. It has anticoagulant property.
Application
Biochem/physiol Actions
Still not finding the right product?
Explore all of our products under Ethylenediaminetetraacetic acid tetrasodium salt dihydrate
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - STOT RE 2
target_organs
Respiratory Tract
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


