429759
Calcium chloride
AnhydroBeads™, −10 mesh, ≥99.99% trace metals basis
Synonym(s):
Calcium dichloride
About This Item
Recommended Products
vapor pressure
0.01 mmHg ( 20 °C)
Quality Level
product line
AnhydroBeads™
Assay
≥99.99% trace metals basis
form
beads
impurities
≤100.0 ppm Trace Metal Analysis
particle size
−10 mesh
mp
772 °C (lit.)
SMILES string
[Cl-].[Cl-].[Ca++]
InChI
1S/Ca.2ClH/h;2*1H/q+2;;/p-2
InChI key
UXVMQQNJUSDDNG-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
accessory
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service