Skip to Content
Merck
All Photos(1)

Key Documents

202940

Sigma-Aldrich

Calcium chloride hydrate

99.999% trace metals basis

Synonym(s):

Calcium dichloride hydrate, Dichlorocalcium hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCl2 · xH2O
CAS Number:
Molecular Weight:
110.98 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.01 mmHg ( 20 °C)

Quality Level

Assay

99.999% trace metals basis

form

crystals
powder and chunks

impurities

≤15.0 ppm Trace Metal Analysis

density

2.15 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.H2O/h;2*1H;1H2/q+2;;;/p-2

InChI key

YMIFCOGYMQTQBP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taisuke Fukawa et al.
Foot & ankle international, 38(6), 596-604 (2017-04-13)
An intra-articular injection of platelet-rich plasma (PRP) may be an effective treatment for osteoarthritis (OA). However, its efficacy in ankle OA has not been investigated yet. The purpose of this study was to assess the safety and efficacy of an
Alessandra Adrover et al.
Pharmaceutics, 11(4) (2019-04-20)
In this study, gellan gum (GG), a natural polysaccharide, was used to fabricate spherical porous beads suitable as sustained drug delivery systems for oral administration. GG was cross-linked with calcium ions to prepare polymeric beads. Rheological studies and preliminary experiments
Marcin Mikulewicz et al.
Biological trace element research, 146(2), 272-280 (2011-10-21)
In this paper, we report the results of an in vitro experiment on the release of metal ions from orthodontic appliances composed of alloys containing iron, chromium, nickel, silicon, and molybdenum into artificial saliva. The concentrations of magnesium, aluminum, silicon
Tatiana V Vygodina et al.
Biochimie, 149, 71-78 (2018-04-11)
Subunit I of cytochrome c oxidase (CcO) from mitochondria and many bacteria contains a cation binding site (CBS) located at the outer positively charged aqueous phase not far from heme a. Binding of Ca2+ with the CBS in bovine CcO
Marie Dubus et al.
Biomaterials science, 8(20), 5763-5773 (2020-09-19)
A multifunctional material system that kills bacteria and drives bone healing is urgently sought to improve bone prosthesis. Herein, the osteoinductive coating made of calcium phosphate/chitosan/hyaluronic acid, named Hybrid, was proposed as an antibacterial substrate for stromal cell adhesion. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service