Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
NACRES:
NA.71
PubChem Substance ID:
UNSPSC Code:
12352302
EC Number:
233-140-8
MDL number:
Assay:
≥99.0%
Form:
powder
Solubility:
H2O: 294 mg/mL, clear, colorless
product line
BioReagent
Quality Level
assay
≥99.0%
form
powder
technique(s)
cell culture | insect: suitable, cell culture | mammalian: suitable, cell culture | plant: suitable
mp
176 °C (dec.) (lit.)
solubility
H2O: 294 mg/mL, clear, colorless
SMILES string
O.O.Cl[Ca]Cl
InChI
1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2
InChI key
LLSDKQJKOVVTOJ-UHFFFAOYSA-L
General description
Calcium chloride (CaCl2) is a commonly used reagent in biochemistry. It plays important roles in many biological processes, including signal transduction, muscle contraction, and maintenance of cell membrane and cell wall stability. CaCl2 is used in the preparation and transformation of competent Escherichia coli (E. coli) and the transfection of eukaryotic cells with either plasmid DNA or high molecular weight genomic DNA. The CaCl2-mediated electroporation of E. coli with the plasmid DNA pBR322 has been studied. CaCl2 is used in the stabilization and two-dimensional crystallization of the NADH-ubiquinone oxidoreductase (complex I) from E. coli. The crystallization of porcine pancreatic elastase in the presence of CaCl2 along with sodium citrate reveals the binding of calcium in the metal-binding site of the protein. The effect of CaCl2 on the total fluorescence in the polymerization of the tubulin-like filamenting temperature-sensitive mutant Z (FtsZ) division protein of E. coli has been studied by a multiwell fluorescent assay.
Application
Calcium chloride dihydrate has been used:
- in the formulation of media
- in sperm collection and preparation
- in cell culture and protoplast preparation
Still not finding the right product?
Explore all of our products under Calcium chloride dihydrate
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
13 - Non Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Product Information Sheet
Hemolymph analysis and evaluation of newly formulated media for culture of shrimp cells (Penaeus stylirostris)
Shimizu C, et al.
In Vitro Cellular & Developmental Biology. Animal, 37(6), 322-329 (2001)
Biomolecule delivery into canola protoplasts by centrifuging cells with microbubbles
He C, et al.
Febs Letters, 587(3), 285-290 (2013)
The effects of L-ergothioneine and L-ascorbic acid on the in vitro maturation (IVM) and embryonic development (IVC) of sheep oocytes
PANCARCI S M, et al.
Kafkas U?niversitesi veteriner faku?ltesi dergisi, 16(5) (2010)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C7902-500G | 04061835543540 |
| C7902-1KG | 04061835543533 |
