Skip to Content
Merck
All Photos(1)

Key Documents

D204307

Sigma-Aldrich

Diphenylacetic acid

99%

Synonym(s):

α,α-Diphenylacetic acid, α-Phenylbenzeneacetic acid, 2,2-Diphenylacetic acid, Diphenylethanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CHCO2H
CAS Number:
Molecular Weight:
212.24
Beilstein:
1910978
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

146-149 °C
147-149 °C (lit.)

SMILES string

OC(=O)C(c1ccccc1)c2ccccc2

InChI

1S/C14H12O2/c15-14(16)13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,13H,(H,15,16)

InChI key

PYHXGXCGESYPCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diphenylacetic acid can be used as a reagent in the kinetic resolution of racemic 2-hydroxy-γ-butyrolactones and 1-heteroarylalkanols by asymmetric esterification in the presence of pivalic anhydride and chiral acyl-transfer catalyst.
It can also be used as:
  • An additive in the ortho-arylation of 1-phenyl-β-carbolines using various aryl halides in the presence of ruthenium catalyst.
  • A catalyst to synthesize 2-allyl-3-oxazolin-5-one derivatives via Rh-catalyzed coupling reaction of azlactones and alkynes followed by aza-Cope rearrangement.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

383.5 °F - closed cup

Flash Point(C)

195.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subramani Rajkumar et al.
The Journal of organic chemistry, 80(11), 5532-5545 (2015-04-11)
A Ru(II)-catalyzed C-H arylation approach has been developed utilizing β-carboline alkaloids as the directing group. Selective formations of diarylated products from moderate to excellent yields were accomplished. Broad substrate scope with excellent functional group tolerance for C1-phenyl/thienyl/PAHs-β-carbolines was demonstrated. X-ray
Jinqiang Kuang et al.
Angewandte Chemie (International ed. in English), 56(29), 8422-8425 (2017-05-18)
Rhodium-catalyzed regioselective addition of azlactones to internal alkynes combined with aza-Cope rearrangement provides efficient atom economic access to 2-allyl-3-oxazolin-5-one derivatives. Extension to a triple domino process, in which the above process is combined with in situ azlactone formation starting from amino
Kinetic Resolution of Racemic 1-Heteroarylalkanols by Asymmetric Esterification Using Diphenylacetic Acid with Pivalic Anhydride and a Chiral Acyl-transfer Catalyst
Shiina I, et al.
Chemistry Letters (Jpn), 40(2), 147-149 (2011)
Hugh D Glossop et al.
Biomacromolecules, 20(7), 2515-2529 (2019-05-31)
Peptides can serve as versatile therapeutics with a highly modular structure and tunable biophysical properties. In particular, the efficacy of peptide antibiotics against drug-resistant pathogens is of great promise, as few new classes of antibiotics are being developed to overcome
B C Sallustio et al.
The Journal of pharmacology and experimental therapeutics, 251(1), 288-294 (1989-10-01)
Drugs which are cleared predominantly by forming acyl-glucuronides undergo a futile cycle in which plasma drug clearance is a function of the formation, hydrolysis and renal clearance of the glucuronide conjugate. The effect of impaired renal function, induced by uranyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service