40410
N,N′-Dimethylurea
(sym.), ≥95.0% (HPLC), technical
Synonym(s):
1,3-Dimethylurea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3NH)2CO
CAS Number:
Molecular Weight:
88.11
Beilstein:
1740672
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
grade
technical
Assay
≥95.0% (HPLC)
bp
268-270 °C (lit.)
mp
101-104 °C (lit.)
102-109 °C
functional group
amine
SMILES string
CNC(=O)NC
InChI
1S/C3H8N2O/c1-4-3(6)5-2/h1-2H3,(H2,4,5,6)
InChI key
MGJKQDOBUOMPEZ-UHFFFAOYSA-N
Gene Information
human ... EPHX2(2053)
mouse ... Ephx2(13850)
Looking for similar products? Visit Product Comparison Guide
General description
N,N′-Dimethylurea (1,3-Dimethylurea), an alkyl urea derivative, is a nonlinear organic material. It forms needle-shaped crystals. It has been crystallized by using ethylacetate (solvent) and heptane (precipitant) by vapor diffusion technique. Its crystals has one molecule in each asymmetric unit. Molecules in crystal are linked by hydrogen bonds.
Application
N,N′-Dimethylurea is the suitable reagent used to investigate the polar structure of its crystals. It may be used in the Dowex-50W ion exchange resin-promoted solvent-free synthesis of N,N′-disubstituted-4-aryl-3,4-dihydropyrimidinones.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
314.6 °F - closed cup
Flash Point(C)
157 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Infrared spectra and configurations of alkylurea derivatives: Normal vibrations on N,N'-dimethyl-and tetramethylurea.
Rao CNR, et al.
Journal of Molecular Spectroscopy, 28(4), 526-535 (1968)
Nonlinear optical properties of N,N' dimethylurea.
Halbout JM, et al.
Applied Physics Letters, 37(10), 864-866 (1980)
Polar structure of N,N'-dimethylurea crystals.
Perez-Folch J, et al.
Journal of Chemical Crystallography, 27(6), 367-369 (1997)
Dowex-promoted general synthesis of N, N'-disubstituted-4-aryl-3, 4-dihydropyrimidinones using a solvent-free Biginelli condensation protocol.
Singh K, et al.
Tetrahedron Letters, 47(25), 4205-4207 (2006)
Anna Ronowicz et al.
Analytical biochemistry, 426(2), 91-93 (2012-04-17)
Reuse of materials in DNA hybridization-based methods has been known since the advent of Southern membranes. Array-based comparative genomic hybridization is essentially Southern hybridization with multiple probes immobilized on a solid surface. We show that comparative genomic hybridization microarrays fabricated
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service