Skip to Content
Merck

8.03244

N,N′-Dimethylurea

for synthesis

Synonym(s):

N,N′-Dimethylurea, 1,3-Dimethylurea

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C3H8N2O
CAS Number:
Molecular Weight:
88.11
UNSPSC Code:
12352111
EC Index Number:
202-498-7
NACRES:
NA.22
MDL number:
Form:
crystals
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

6 hPa ( 115 °C)

Quality Segment

form

crystals

autoignition temp.

400 °C

potency

4000 mg/kg LD50, oral (Rat)

pH

9.0-9.5 (20 °C, 100 g/L in H2O)

bp

268-270 °C/1013 hPa

mp

105-108 °C

transition temp

flash point 154 °C (DIN 51758)

density

1.14 g/cm3 at 20 °C

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

N(C)C(=O)NC

InChI

1S/C3H8N2O/c1-4-3(6)5-2/h1-2H3,(H2,4,5,6)

InChI key

MGJKQDOBUOMPEZ-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Melting range (lower value): ≥ 102 °C
Melting range (upper value): ≤ 107 °C
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under N,N′-Dimethylurea


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

314.6 °F - closed cup

flash_point_c

157 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library