Skip to Content
Merck

36574

N,N′-Dimethylurea

PESTANAL®, analytical standard

Synonym(s):

1,3-Dimethylurea

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3NH)2CO
CAS Number:
Molecular Weight:
88.11
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
202-498-7
Beilstein/REAXYS Number:
1740672
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

description

(sym.)

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

bp

268-270 °C (lit.)

mp

101-104 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)NC

InChI

1S/C3H8N2O/c1-4-3(6)5-2/h1-2H3,(H2,4,5,6)

InChI key

MGJKQDOBUOMPEZ-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under N,N′-Dimethylurea


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

314.6 °F - closed cup

flash_point_c

157 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library