426466
Methylamine solution
40 wt. % in H2O
Synonym(s):
Monomethylamine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
CH3NH2
CAS Number:
Molecular Weight:
31.06
Beilstein:
741851
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
bp:
48 °C
vapor pressure:
280 mmHg ( 20 °C)
659 mmHg ( 40 °C)
659 mmHg ( 40 °C)
Recommended Products
vapor density
1.07 (vs air)
Quality Level
vapor pressure
280 mmHg ( 20 °C)
659 mmHg ( 40 °C)
autoignition temp.
806 °F
concentration
40 wt. % in H2O
refractive index
n20/D 1.37
bp
48 °C
density
0.897 g/mL at 25 °C
functional group
amine
SMILES string
CN
InChI
1S/CH5N/c1-2/h2H2,1H3
InChI key
BAVYZALUXZFZLV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Methylamine solution may be used as a LC-MS/MS mobile phase in the perchlorate analysis of milk and yogurt samples.
It may be used in the synthesis of the following:
It may be used in the synthesis of the following:
- 9-fluorenylmethyl N-methylcarbamate and tert-butyl N-methylcarbamate
- 7-methylamino-2-(2-quinolyl)tropone
- 2,5-meso-pyrrolidines
- 2-(4-chlorophenyl)-N-methylacetamide
- 1,3-benzoxazine derivatives from eugenol
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
14.0 °F - closed cup
Flash Point(C)
-10 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
An improved scalable route to pure dronedarone hydrochloride.
Hivarekar RR, et al.
Organic Process Research & Development, 16(4), 677-681 (2012)
Chemical profiling of seized methamphetamine putatively synthesized from phenylacetic acid derivatives.
Tsujikawa K, et al.
Forensic Science International, 227(1), 42-44 (2013)
Synthesis and brine shrimp lethality test of some benzoxazine and aminomethyl derivatives of eugenol.
Rudyanto M, et al.
International Journal of Pharmacy and Pharmaceutical Sciences, 6, 96-98 (2014)
Orthogonally Protected N-Methyl-Substituted α-Aminoglycines.
Jiang G, et al.
Protein and peptide letters, 3(4), 219-224 (1996)
Preparation of Tetrahydroisoquinoline-3-ones Via Cyclization of Phenyl Acetamides Using Eaton's Reagent.
Yang Q, et al.
Organic Syntheses, 44-54 (2012)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service