Skip to Content
Merck
All Photos(1)

Documents

394912

Supelco

Heptafluorobutyric anhydride

for GC derivatization, LiChropur, ≥99.0%

Synonym(s):

HFBA, HFAA, Perfluorobutyric anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3CF2CF2CO)2O
CAS Number:
Molecular Weight:
410.06
Beilstein:
856036
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Assay

≥99.0%

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.287 (lit.)

bp

108-110 °C (lit.)

mp

−43 °C (lit.)

density

1.674 g/mL at 20 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C8F14O3/c9-3(10,5(13,14)7(17,18)19)1(23)25-2(24)4(11,12)6(15,16)8(20,21)22

InChI key

UFFSXJKVKBQEHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heptafluorobutyric anhydride (HFB) is a derivatizing reagent. It provides facile and sensitive way for detecting and elucidating structure of numerous alkyl-chain homologs.

Application

Learn more in the Product Information
GC derivatizing agent used to prepare volatile derivatives of amino acids and other biologically active compounds.
HFB was used as derivatizing agent in analysing platelet-activating factor using GC-MS.
Suitable for the derivatization of aldosterone, digoxin, digitoxin and metabolites, estradiol, postaglandins F and F2α, 3-oxo-Δ4-steroids, and testosterone.

Packaging

Derivatization reagent used in a GC-MS analysis of stereoisomer content of synthetic peptides.

Other Notes

Reagent for heptafluorobutyrate, 3-enol heptafluorobutyrate, 17-heptafluorobutyrate, heptafluorobutyrate and heptafluorobutyryl.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S T Weintraub et al.
Journal of lipid research, 31(4), 719-725 (1990-04-01)
Parallel analysis of platelet-activating factor (PAF) using chemical ionization gas chromatography-mass spectrometry after direct derivatization with pentafluorobenzoyl chloride (PFB) and heptafluorobutyric anhydride (HFB) provides a facile and highly sensitive means for detecting and elucidating the structure of the numerous alkyl-chain
J Bakthavachalam et al.
Journal of chromatography, 500, 373-386 (1990-02-02)
Several electrophoric derivatives of 1-aminopyrene and 2-aminofluorene were prepared. Reagents such as heptafluorobutyryl chloride, pentafluorobenzoyl chloride, pentafluorobenzyl bromide and pentafluorobenzaldehyde, alone and in certain combinations, were employed. The ease of formation, yield, stability and fragmentation by gas chromatography with electron-capture
Ralf Pätzold et al.
Chirality, 18(7), 551-557 (2006-05-10)
Synthetic dipeptides comprising mixtures of enantiomers, diastereomers, or sequential isomers were converted into their N-perfluoroacetyl dipeptide esters (perfluoroacetyl: trifluoroacetyl, pentafluoroacetyl, heptafluorobutyryl; ester: methyl, 1-propyl, 2-propyl, 2,2,2-trifluoroethyl) and analyzed by GC-MS on the chiral stationary phases Chirasil-L-Val and Lipodex-E using helium
Ronald Agius et al.
Forensic science international, 196(1-3), 3-9 (2010-01-12)
The analysis of ethyl glucuronide (EtG) in hair is a powerful tool for chronic alcohol abuse control because of the typical wide detection window of the hair matrix and due to the possibility of segmentation, allowing evaluation of alcohol consumption
O Beck et al.
Journal of analytical toxicology, 22(1), 45-49 (1998-03-10)
The use of mushrooms containing the hallucinogenic substance psilocybin for intentional intoxication is relatively common. Occasionally, this results in adverse reactions with typical tachycardia that is not evidently caused by psilocybin. This study demonstrates the presence of phenylethylamine in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service