Skip to Content
Merck

77249

Heptafluorobutyric acid

≥99.0% (GC)

Synonym(s):

Edman Reagent No. 3, HFBA, Perfluorobutyric acid

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
CF3CF2CF2COOH
CAS Number:
Molecular Weight:
214.04
UNSPSC Code:
12352106
NACRES:
NA.26
PubChem Substance ID:
EC Number:
206-786-3
Beilstein/REAXYS Number:
1426882
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

vapor density

7 (vs air)

Quality Level

vapor pressure

~10 mmHg ( 25 °C)

assay

≥99.0% (GC)

form

liquid

analyte chemical class(es)

amino acids

refractive index

n20/D 1.3 (lit.)

bp

120 °C/755 mmHg (lit.)

density

1.645 g/mL at 25 °C (lit.)

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C4HF7O2/c5-2(6,1(12)13)3(7,8)4(9,10)11/h(H,12,13)

InChI key

YPJUNDFVDDCYIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

In protein sequencing with converted Beckman gas-liquid phase sequencer.

signalword

Danger

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter

pictograms

CorrosionExclamation mark

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A S Bhown et al.
Analytical biochemistry, 175(1), 39-51 (1988-11-15)
As an effective aid to extend the microsequencing capabilities the Beckman protein/peptide sequenator Series 890C has been successfully converted to a gas-liquid system, in which coupling buffer 25% trimethylamine was employed as a gas, and heptafluorobutyric acid as a liquid.
Ulrika Eriksson et al.
Environmental science and pollution research international, 20(11), 7940-7948 (2013-04-17)
Diet and drinking water are suggested to be major exposure pathways for perfluoroalkyl substances (PFASs). In this study, food items and water from Faroe Islands sampled in 2011/2012 were analyzed for 11 perfluoroalkyl carboxylic acids (PFCAs) and 4 perfluoroalkane sulfonic
Yuko Murakami et al.
Organic & biomolecular chemistry, 12(48), 9887-9894 (2014-10-31)
Desmosine-CH2, an analog of the elastic tissue degradation biomarker desmosine, can be regarded as a potential internal standard for precise quantification of desmosines by LC-MS/MS. In this study, the chemical synthesis of desmosine-CH2 was completed in 22% overall yield in
V Gellrich et al.
Chemosphere, 87(9), 1052-1056 (2012-03-07)
Perfluorinated compounds (PFCs) can be detected worldwide in both, soil and water. In order to study the leaching behavior of this heterogeneous group of compounds in soil, flow-through column experiments have been conducted. Ten perfluoro carboxylates and four perfluoro sulfonates
Kaberi P Das et al.
Toxicological sciences : an official journal of the Society of Toxicology, 105(1), 173-181 (2008-05-31)
Perfluorobutyrate (PFBA) is a perfluoroalkyl acid (PFAA) found in the environment. Previous studies have indicated developmental toxicity of PFAAs (perfluorooctane sulfonate [PFOS] and perfluorooctanoate [PFOA]); the current study examines that of PFBA. PFBA/NH4(+) was given to timed-pregnant CD-1 mice by

Global Trade Item Number

SKUGTIN
77249-10ML04061837839221
77249-50ML04061832927060

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service