Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Heptafluorobutyric anhydride
Synonym(s):
HFAA, HFBA, Perfluorobutyric anhydride
Linear Formula:
(CF
3
CF
2
CF
2
CO)
2
O
CAS Number:
336-59-4
Molecular Weight:
410.06
EC Number:
206-410-8
Products
Product Category
building blocks (2)
Brand
Supelco (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Special Grade
Puriss (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
H1006
Heptafluorobutyric anhydride,
≥98%
Expand
View Pricing
394912
Heptafluorobutyric anhydride,
derivatization grade (GC derivatization), LiChropur
™
, ≥99.0%
Expand
View Pricing
77253
Heptafluorobutyric anhydride,
derivatization grade (GC derivatization), LiChropur
™
, ≥99.0%
Expand
View Pricing