Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
derivatization grade (GC derivatization)
Quality Segment
assay
≥99.0% (GC), ≥99.0%
form
liquid
quality
LiChropur™
reaction suitability
reagent type: derivatization reagent
reaction type: Acylations
technique(s)
gas chromatography (GC): suitable
refractive index
n20/D 1.287 (lit.)
bp
108-110 °C (lit.)
mp
−43 °C (lit.)
density
1.674 g/mL at 20 °C
SMILES string
FC(F)(F)C(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)C(F)(F)F
InChI
1S/C8F14O3/c9-3(10,5(13,14)7(17,18)19)1(23)25-2(24)4(11,12)6(15,16)8(20,21)22
InChI key
UFFSXJKVKBQEHC-UHFFFAOYSA-N
General description
Application
Packaging
Analysis Note
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥99.0% (GC) | assay ≥99.0% | assay ≥98% | assay ≥99.0% (GC) |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| reaction suitability reagent type: derivatization reagent | reaction suitability reagent type: derivatization reagent | reaction suitability - | reaction suitability reagent type: derivatization reagent |
| bp 108-110 °C (lit.) | bp 108-110 °C (lit.) | bp 108-110 °C (lit.) | bp 39.5-40 °C (lit.) |
| mp −43 °C (lit.) | mp −43 °C (lit.) | mp −43 °C (lit.) | mp −65 °C (lit.) |
| density 1.674 g/mL at 20 °C | density 1.674 g/mL at 20 °C (lit.) | density 1.674 g/mL at 20 °C (lit.) | density 1.511 g/mL at 20 °C (lit.) |
Still not finding the right product?
Explore all of our products under Heptafluorobutyric anhydride
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


