Skip to Content
Merck
All Photos(2)

Documents

387932

Sigma-Aldrich

Poly(vinyl acetate)

average Mw ~500,000 by GPC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(O2CCH3)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

beads

Quality Level

mol wt

average Mw ~500,000 by GPC

storage temp.

2-8°C

SMILES string

COC(=O)C=C

InChI

1S/C4H6O2/c1-3-6-4(2)5/h3H,1H2,2H3

InChI key

XTXRWKRVRITETP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Andonova et al.
Die Pharmazie, 67(7), 601-604 (2012-08-15)
During the last decade the number of investigations on the preparation and application of more effective drug release systems on the basis of nanocarriers from biocompatible and biodegradable polymers are considerably increasing. This is notably in force for practically water
Vibha Puri et al.
Journal of pharmaceutical sciences, 101(1), 342-353 (2011-09-22)
Amorphous solid dispersions (ASDs) may entail tailor-made dosage form design to exploit their solubility advantage. Surface phenomena dominated the performance of amorphous celecoxib solid dispersion (ACSD) comprising of amorphous celecoxib (A-CLB), polyvinylpyrrolidone, and meglumine (7:2:1, w/w). ACSD cohesive interfacial interactions
Il Gyu Kang et al.
American journal of rhinology & allergy, 24(5), 392-395 (2011-01-20)
The aim of this study was to evaluate the effect of repeated expandable polyvinyl acetate (EPA) packing for preventing stenosis of the frontal sinus ostium after patients undergo functional endoscopic sinus surgery (FESS). A nonrandomized, prospective study of 20 patients
Seon Tae Kim et al.
International journal of pediatric otorhinolaryngology, 77(1), 113-116 (2012-11-08)
The purpose of this prospective study was to determine the effectiveness of polyurethane foam (PUF) and polyvinyl acetate (PA) as packing materials for reducing post-conchotomy bleeding, pain, and headaches. This study was a prospective, randomized and single-blinded controlled study. Fifty-two
Dae Woo Kim et al.
American journal of rhinology & allergy, 26(5), e147-e149 (2012-11-22)
This study investigated the efficacy of glove finger-coated polyvinyl acetate (PA) pack on hemostasis, pain levels, and wound healing after endoscopic sinus surgery (ESS). A prospective, randomized, double-blinded controlled study was performed in 30 patients who underwent bilateral ESS for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service