Skip to Content
Merck
All Photos(1)

Documents

81388

Sigma-Aldrich

Poly(vinyl chloride)

low molecular weight

Synonym(s):

PVC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CHCl)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

K-value 55 - 57

form

powder

mol wt

~48000

density

1.4 g/mL at 25 °C (lit.)

SMILES string

ClC=C

InChI

1S/C2H3Cl/c1-2-3/h2H,1H2

InChI key

BZHJMEDXRYGGRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(vinyl chloride) (PVC) is a commercially sold thermoplastic material with excellent physical properties. It can be used for a variety of applications such as water purification and artificial muscle structure.

Application


  • Environmental Stability of PVC: Study on the environmental stability of poly(vinyl chloride) modified by Schiff′s base, highlighting its resistance to UV light (Ahmed et al., 2021).

  • Mechanical Properties of PVC Nanofibers: Research on the effects of polymer concentration on the morphology and mechanical properties of electrospun cellulose acetate and PVC nanofiber mats (Tarus et al., 2016).

  • Photostabilization of PVC: Insights into the ongoing advancements in the photostabilization of PVC to enhance its longevity and material stability (Yousif & Hasan, 2015).

  • PVC and Mn2+ Doped Films: Investigation of the structural, thermal, and optical properties of pure and Mn2+ doped PVC films, exploring material enhancements (Kayyarapu et al., 2016).

  • PVC Plasticizer Migration: A review on methods for determination of plasticizer migration from PVC materials, addressing implications for durability and material safety (Chen et al., 2022).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of Ultralow Molecular Weight Poly (vinyl chloride) with Submicrometer Particles via Precipitation Polymerization
Cui P, et al.
Chinese Journal of Polymer Science, 24(3), 1-8 (2005)
Effect of hydrophobicity of polymer materials used for water purification membranes on biofilm formation dynamics
Saeki D, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 506(3), 622-628 (2016)
Poly (vinyl chloride)(PVC) hollow fiber ultrafiltration membranes prepared from PVC/additives/solvent
Xu J and Xu Z
Journal of Membrane Science , 208(1-2), 203-212 (2002)
Towards electroactive gel artificial muscle structures
Electroactive Polymer Actuators and Devices (EAPAD) XX, 10594(1-2), 1059408-1059408 (2018)
Hiroyuki Yanagisawa et al.
Molecules (Basel, Switzerland), 23(4) (2018-03-23)
This study was conducted with the aim of achieving the simultaneous screening of various additives in polymer materials by utilizing a solvent-free pyrolyzer/thermal desorption gas chromatography mass spectrometry (Py/TD-GC-MS) method. As a first step to achieve this goal, simultaneous screening

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service