Skip to Content
MilliporeSigma

Skip To

190845

Poly(1-vinylpyrrolidone-co-vinyl acetate)

average Mw ~50,000 (GPC vs. poly(ethylene oxide)), powder

Synonym(s):

PVP-VA copolymer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 g

Available to ship TODAYfromMILWAUKEE

$214.00

About This Item

CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23
MDL number:

$214.00


Available to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

mol wt

average Mw ~50,000 (GPC vs. poly(ethylene oxide))

transition temp

Tg 64 °C

density

1.27 g/mL at 25 °C (lit.)

SMILES string

N1(CCCC1=O)C=C.O(C(=O)C)C=C

InChI

1S/C6H9NO.C4H6O2/c1-2-7-5-3-4-6(7)8;1-3-6-4(2)5/h2H,1,3-5H2;3H,1H2,2H3

InChI key

FYUWIEKAVLOHSE-UHFFFAOYSA-N

Application

Poly(1-vinylpyrrolidone-co-vinyl acetate) can be used as a starting material to prepare a quasi-solid polymer electrolyte (QSPE) for dye-sensitized solar cells.

It can be used to preparepolymer blend membranes for pervaporation separation of ethanol and2-ethylhexanol mixtures.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
188050416320416339
form

powder

form

powder

form

powder

form

powder

density

1.27 g/mL at 25 °C (lit.)

density

-

density

-

density

-

mol wt

average Mw ~50,000 (GPC vs. poly(ethylene oxide))

mol wt

average Mw 100,000-500,000

mol wt

average Mn ~311,000, average Mw ~1,080,000

mol wt

average Mn ~80,000, average Mw ~216,000

transition temp

Tg 64 °C

transition temp

Tg 235 °C

transition temp

-

transition temp

-

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

100


Storage Class

11 - Combustible Solids

wgk

WGK 1

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is product 190845 a block copolymer or a random copolymer? Additionally, what is the Vinyl Acetate content in the polymer?

    1 answer
    1. This copolymer is a linear, random copolymer produced by the free-radical polymerization of vinyl acetate (VA) to vinylpyrrolidone (VP). The polymer has no free vinyl acetate present, but the ratio of polymer vinylpyrrolidone to polymer vinyl acetate is 6:4.

      Helpful?

Reviews

No rating value

Active Filters