Skip to Content
Merck
All Photos(2)

Documents

340502

Sigma-Aldrich

Poly(ethylene-co-vinyl acetate)

vinyl acetate 40 wt. %, melt index (41-63 dg/min (190°C/2.16kg)), contains 190-910 ppm inhibitor

Synonym(s):

PEVA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH2)m[CH2CH(OCOCH3)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

beads

autoignition temp.

500 °F

melt index

(41-63 dg/min (190°C/2.16kg))

contains

190-910 ppm inhibitor

composition

vinyl acetate, 40 wt. %

hardness

40 (Shore A-2, ASTM D 2240)

transition temp

Tg −40-−30 °C
Tm 110-120 °C

solubility

toluene, THF, and MEK: soluble

SMILES string

C=C.CC(=O)OC=C

InChI

1S/C4H6O2.C2H4/c1-2-3-4(5)6;1-2/h2H,1,3H2,(H,5,6);1-2H2

InChI key

DQXBYHZEEUGOBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene-co-vinyl acetate) (PEVA) is a flame-retardant material with good mechanical and physical properties. It is majorly used as an insulating material in wire and cable industry.

Application

  • Correlation between Cohesive Energy Density, Fractional Free Volume, and Gas Transport Properties: This study explores the properties of EVA materials that affect their gas permeability, important for applications like packaging and barrier materials (Kubica & Wolinska-Grabczyk, 2015).
  • The Thermal and Mechanical Properties of EVA and its Crosslinked Analogues: This research investigates how crosslinking affects the thermal and mechanical properties of EVA, which could interest material scientists working with polymers (Wang & Deng, 2019).
  • Investigation of morphological, rheological, and mechanical properties of cyclic olefin copolymer/EVA blend films: This study focuses on the blend properties of EVA with other polymers, providing insights useful in academia and material science (Durmus et al., 2018).
  • Fabrication, characterization, and properties of EVA/magnetite nanocomposites: Relevant to material scientists, this research discusses the incorporation of magnetite nanoparticles into EVA for enhanced properties (Ramesan, 2014).

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly (ethylene-co-vinyl acetate)/clay nanocomposites: Effect of clay nature and organic modifiers on morphology, mechanical and thermal properties
Peeterbroeck S, et al.
Polymer Degradation and Stability, 90(2), 288-294 (2005)
Preparation and flammability of a novel intumescent flame-retardant poly (ethylene-co-vinyl acetate) system
Wang D, et al.
Polymer Degradation and Stability, 93(12), 2186-2192 (2008)
Lili Wang et al.
PloS one, 7(6), e37781-e37781 (2012-06-14)
The study on the rare earth (RE)-doped layered double hydroxides (LDHs) has received considerable attention due to their potential applications in catalysts. However, the use of RE-doped LDHs as polymer halogen-free flame retardants was seldom investigated. Furthermore, the effect of
Neide Pena Coto et al.
Dental traumatology : official publication of International Association for Dental Traumatology, 28(2), 108-113 (2011-07-28)
There has been a significant increase in the number of facial fractures stemming from sport activities in recent years, with the nasal bone one of the most affected structures. Researchers recommend the use of a nose protector, but there is
Taiji Ogawa et al.
International journal of oral science, 4(1), 14-18 (2012-03-17)
Bacterial infection associated with the use of medical or dental devices is a serious concern. Although devices made of ethylene vinyl acetate (EVA) are often used in the oral cavity, there are no established standards for their storage. We investigated

Articles

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service