Skip to Content
Merck
All Photos(2)

Key Documents

359025

Sigma-Aldrich

Didodecyldimethylammonium bromide

98%

Synonym(s):

DDAB, Dimethyldidodecylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)11]2N(CH3)2(Br)
CAS Number:
Molecular Weight:
462.63
Beilstein:
3576662
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

157-162 °C (lit.)

SMILES string

[Br-].CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC

InChI

1S/C26H56N.BrH/c1-5-7-9-11-13-15-17-19-21-23-25-27(3,4)26-24-22-20-18-16-14-12-10-8-6-2;/h5-26H2,1-4H3;1H/q+1;/p-1

InChI key

XRWMGCFJVKDVMD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Didodecyldimethylammonium bromide (DDAB) is a double-chain cationic surfactant.

Possible applications:
  • DDAB can be employed as a co-surfactant to synthesize well-defined multilamellar vesicular silica with an adjustable number of layers.
  • As a surfactant to synthesize gold nanoclusters and nanocubes.
  • It forms supersaturated reverse micelles and microemulsions that act as organized reaction microenvironments for the synthesis of barium sulfate.
  • Calcium montmorillonite can be modified by DDAB through direct ion-exchange to synthesize thermally stable organoclay.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of block length and structure of surfactant on self-assembly and solution behavior of block ionomer complexes.
Bronich TK.
Langmuir, 16(2), 481-489 (2000)
A cationic-cationic co-surfactant templating route for synthesizing well-defined multilamellar vesicular silica with an adjustable number of layers.
Zhang Y.
Chemical Communications (Cambridge, England), 50(22), 2907-2909 (2014)
R A Carvalho et al.
Journal of colloid and interface science, 354(2), 725-732 (2010-12-15)
NMR spectroscopy has been used to study and characterize the interactions in solution between β-CD and alkyl-α,ω-bis(dodecyldimethyl ammonium bromide) gemini surfactants with the following head-group spacer lengths: 2, 4, 6, 8, and 10. The application of the method of continuous
Sana Ghrab et al.
Environmental science and pollution research international, 25(19), 18251-18262 (2017-05-14)
Essential oils from aromatic plants are currently mentioned as suitable tools for excellent protection of stored grains from insect pest attacks. The present work aimed to study the processes of the synthesis insecticidal formulation with clay. The active terpenic compounds
Thermally-induced formation of atomic Au clusters and conversion into nanocubes.
Jin R.
Journal of the American Chemical Society, 126(32), 9900-9901 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service