Skip to Content
Merck

Skip To

B5776

Benzyldodecyldimethylammonium bromide

Synonym(s):

Benzyldimethyldodecylammonium bromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)11N(Br)(CH3)2CH2C6H5
CAS Number:
Molecular Weight:
384.44
UNSPSC Code:
12161900
NACRES:
NA.25
PubChem Substance ID:
EC Number:
230-698-4
Beilstein/REAXYS Number:
3579193
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

cationic

Quality Segment

assay

≥96.5% (TLC)

form

powder

mol wt

384.44 g/mol

storage temp.

2-8°C

SMILES string

[Br-].CCCCCCCCCCCC[N+](C)(C)Cc1ccccc1

InChI

1S/C21H38N.BrH/c1-4-5-6-7-8-9-10-11-12-16-19-22(2,3)20-21-17-14-13-15-18-21;/h13-15,17-18H,4-12,16,19-20H2,1-3H3;1H/q+1;/p-1

InChI key

KHSLHYAUZSPBIU-UHFFFAOYSA-M

General description

Benzyldodecyldimethylammonium bromide is a cationic surfactant that may be used as an adsorbent for certain metals such as Cr(VI).

Application

Benzyldodecyldimethylammonium bromide has been used in a study to assess the relation between chemical structure and germicidal efficacy in surface-active cations. It has also been used in a study to investigate pore size control during the synthesis of macroporous thermosensitive poly(N-isopropylacrylamide) hydrogels.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
105098213373C0636
assay

≥96.5% (TLC)

assay

-

assay

≥99.0% (AT)

assay

≥98% (non-aqueous titration)

form

powder

form

-

form

solid

form

powder

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

2-8°C

description

cationic

description

-

description

cationic

description

cationic

mol wt

384.44 g/mol

mol wt

-

mol wt

384.44 g/mol

mol wt

378.47 g/mol

Quality Level

200

Quality Level

-

Quality Level

200

Quality Level

100


Still not finding the right product?


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters