Skip to Content
Merck

C0636

Ethylhexadecyldimethylammonium bromide

≥98% (non-aqueous titration)

Synonym(s):

Cetyldimethylethylammonium bromide, Dimethylethylhexadecylammonium bromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[CH3(CH2)15N+(CH3)2CH2CH3]Br-
CAS Number:
Molecular Weight:
378.47
UNSPSC Code:
12161900
NACRES:
NA.25
PubChem Substance ID:
EC Number:
204-672-8
Beilstein/REAXYS Number:
3729134
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

cationic

Quality Segment

assay

≥98% (non-aqueous titration)

form

powder

mol wt

378.47 g/mol

composition

C16, ~85% (balance stearyl (C18) homolog)

storage temp.

2-8°C

SMILES string

[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)CC

InChI

1S/C20H44N.BrH/c1-5-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(3,4)6-2;/h5-20H2,1-4H3;1H/q+1;/p-1

InChI key

VUFOSBDICLTFMS-UHFFFAOYSA-M

General description

Ethylhexadecyldimethylammonium bromide is a surfactant.

Application

Ethylhexadecyldimethylammonium bromide has been used in a study to assess variables affecting the foam separation of Escherichia coli.


Still not finding the right product?


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library