Skip to Content
Merck
All Photos(2)

Documents

347817

Sigma-Aldrich

Trifluoromethanesulfonic acid

ReagentPlus®, ≥99%

Synonym(s):

Triflic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3H
CAS Number:
Molecular Weight:
150.08
Beilstein:
1812100
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.2 (vs air)

vapor pressure

8 mmHg ( 25 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

refractive index

n20/D 1.327 (lit.)

bp

162 °C (lit.)

density

1.696 g/mL at 25 °C (lit.)

SMILES string

OS(=O)(=O)C(F)(F)F

InChI

1S/CHF3O3S/c2-1(3,4)8(5,6)7/h(H,5,6,7)

InChI key

ITMCEJHCFYSIIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trifluoromethanesulfonic acid is a strong organic acid. It can be prepared by reacting bis(trifluoromethylthio)mercury with H2O2. On mixing with HNO3, it affords a nitrating reagent (a nitronium salt). This reagent is useful for the nitration of aromatic compounds. Its dissociation in various organic solvents has been studied.

Application

Trifluoromethanesulfonic acid can be used as a catalyst to prepare:
  • Substituted tetrahydrofurans and tetrahydropyrans by cyclization of corresponding unsaturated alcohols under acidic conditions.
  • Nitriles from corresponding aldehydes by Schmidt reaction.
  • Disubstituted five-membered ring lactones by allylboration reaction between 2-alkoxycarbonyl allylboronates and aldehydes.

It can also be used as a catalyst in the Fischer glycosylation and Friedel-Crafts acylation reactions.
Deglycosylation agent

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

>332.1 °F - Pensky-Martens closed cup

Flash Point(C)

> 166.7 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of trifluoromethanesulfonic acid in fischer glycosylations.
Wessel HP.
Journal of Carbohydrate Chemistry, 7(1), 263-269 (1988)
Chemoselective Schmidt reaction mediated by triflic acid: selective synthesis of nitriles from aldehydes
Rokade BV and Prabhu KR
The Journal of Organic Chemistry, 77(12), 5364-5370 (2012)
Triflic acid-catalysed cyclisation of unsaturated alcohols
Coulombel L and Dunach E
Green Chemistry, 6(10), 499-501 (2004)
Triflic acid-catalyzed additions of 2-alkoxycarbonyl allylboronates to aldehydes. Study of scope and mechanistic investigation of the reaction stereochemistry
Elford TG, et al.
The Journal of Organic Chemistry, 72(4), 1276-1284 (2007)
Electrochemical studies of sulfonates in non-aqueous solvents: Part III. Trifluoromethanesulfonic acid as a strong acid in dipolar aprotic solvents and acetic acid.
Fujinaga T and Sakamoto I.
J. Electroanal. Chem. Interfac. Electrochem., 85(1), 185-201 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service