Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trifluoromethanesulfonic acid
Synonym(s):
Triflic acid, Trifluoromethanesulfonic acid
Linear Formula:
CF
3
SO
3
H
CAS Number:
1493-13-6
Molecular Weight:
150.08
EC Number:
216-087-5
Products
Product Category
organic acids (2)
acids (1)
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
347817
Trifluoromethanesulfonic acid,
ReagentPlus
®
, ≥99%
Expand
View Pricing
8.21166
Trifluoromethanesulfonic acid,
for synthesis
Expand
View Pricing
158534
Trifluoromethanesulfonic acid,
reagent grade, 98%
Expand
View Pricing