Skip to Content
Merck
All Photos(1)

Key Documents

91737

Sigma-Aldrich

Trifluoromethanesulfonic anhydride

purum, ≥98.0% (T)

Synonym(s):

Tf2O, Triflic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO2)2O
CAS Number:
Molecular Weight:
282.14
Beilstein:
1813600
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.2 (vs air)

Quality Level

vapor pressure

8 mmHg ( 20 °C)

grade

purum

Assay

≥98.0% (T)

form

liquid

refractive index

n20/D 1.321 (lit.)

bp

81-83 °C (lit.)

density

1.677 g/mL at 25 °C (lit.)

functional group

fluoro
triflate

SMILES string

FC(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F

InChI

1S/C2F6O5S2/c3-1(4,5)14(9,10)13-15(11,12)2(6,7)8

InChI key

WJKHJLXJJJATHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for glycosylation for synthesis of polysaccharides

Reagent for stereoselective synthesis of mannosazide methyl uronate donors

Activator for direct glycosylation with anomeric hydroxy sugars
Trifluoromethanesulfonic anhydride can be used in the preparation of azido sugars such as 6-azido fucose. It can also be used as a promoter for the Friedel-Crafts acylation of aromatic compounds with carboxylic acids in the absence of catalyst.
Tf2O in combination with:
  • Diphenyl sulfoxide, forms an efficient promoter system for the activation of thioglycosides for glycosylation reactions.
  • 2-chloropyridine, can be used for secondary amide activation.
  • Potassium iodide, can effectively mediate the deoxygenation of sulfoxides to form sulfides.
  • Hydrogen peroxide, for the selective oxidation of sulfanes to sulf­oxides in the presence of oxidatively sensitive functional groups.

Tf2O can react with:
  • Ketones in the presence of a base to form vinyl triflates.
  • Magnesium alkoxides to form corresponding symmetrical or unsymmetrical ethers.
  • Sodium azide for the in situ preparation of triflyl azide.

Other Notes

Electrophilic trifyl source. Reviews

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Ox. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

not determinedboils before flash

Flash Point(C)

not determinedboils before flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of pyrimidines by direct condensation of amides and nitriles.
Movassaghi M & Hill MD.
Nature Protocols, 2(8), 2018-2018 (2007)
Synthesis, 85-85 (1982)
H2O2/Tf2O System: An efficient oxidizing reagent for selective oxidation of sulfanes.
Khodaei MM, et al.
Synthesis, 2008(11), 1682-1684 (2008)
On the mechanism of the reaction between ketones and trifluoromethanesulfonic anhydride. An improved and convenient method for the preparation of pyrimidines and condensed pyrimidines.
Garcia Martinez A, et al.
The Journal of Organic Chemistry, 57(5), 1627-1630 (1992)
Ching-Tien Lee,
Advanced Synthesis & Catalysis, 351(18), 3139-3142 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service