Skip to Content
Merck
All Photos(1)

Documents

8.21166

Sigma-Aldrich

Trifluoromethanesulfonic acid

for synthesis

Synonym(s):

Trifluoromethanesulfonic acid, Triflic acid, TFMS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHF3O3S
CAS Number:
Molecular Weight:
150.08
MDL number:
UNSPSC Code:
12352106
EC Index Number:
216-087-5
NACRES:
NA.22

vapor pressure

10 hPa ( 55 °C)

Quality Level

form

liquid

potency

1605 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

pH

<1 ( in H2O)

bp

162 °C/1013 hPa

density

1.71 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/2CHF3O3S.Ba/c2*2-1(3,4)8(5,6)7;/h2*(H,5,6,7);/q;;+2/p-2

InChI key

DXJURUJRANOYMX-UHFFFAOYSA-L

General description

Trifluoromethanesulfonic acid (also known as Triflic acid, TFMSA or TfOH) is an organic acid that is commonly used as a reagent in organic synthesis due to its strong Bronsted acidity, finding applications in catalysis, N-protection, dehydration, activation, and addition reactions., TFMSA is also used for global deprotection and cleavage in Boc SPPS.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS

Literature references:
[1] L. R. Subramanian, et al. (2001) Encyclopedia of Reagents for Organic Synthesis.
[2] R. D. Howells & J. D. Mc Cown JD (1977) Chem. Rev., 77, 69.
[3] J. Stewart, J. Young in “Solid Phase Peptide Synthesis”, Pierce Chemical Company, Rockford, 1984.

Application

Recent applications of trifluoromethanesulfonic acid include:
  • A dehydrating agent for the conversion of secondary alcohols to alkene, or the preparation of anhydrides from carboxylic acids.
  • An acid catalyst in the synthesis of carbohydrates and glycosides.
  • An activating agent for the activation of carboxylic acids and can be used for the synthesis of carboxylic acid derivatives such as esters, amides, and anhydrides.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

332.1 °F - Pensky-Martens closed cup

Flash Point(C)

> 166.7 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient p-toluenesulfonic acid-catalyzed alcohol addition or hydration of unsymmetrical arylalkynes
Olivi N, et al.
Synlett, 2004, 2175-2179 (2004)
Solid Phase Peptide Synthesis, Pierce Chemical Company, Rockford, 1984.
J. Stewart, J. Young
Solid Phase Peptide Synthesis (1984)
Trifluoromethanesulfonic acid and derivatives
R. D. Howells & J. D. Mc Cown JD
Chemical Reviews, 77, 69-69 (1977)
Effect of Lewis acid on catalytic dehydration of a chitin-derived sugar alcohol
Sagawa T, et al.
Molecular Catalysis, 111282-111282 (2020)
L. R. Subramanian, et al.
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service