339776
Nickel(II) carbonate hydroxide tetrahydrate
Synonym(s):
Nickel(II) carbonate, basic
About This Item
Recommended Products
grade
for analytical purposes
form
powder
concentration
45-52.5% Ni (With Dimethylglyoxime, gravimetric)
application(s)
battery manufacturing
SMILES string
O.O.O.O.[Ni++].[Ni++].O[Ni]O.O[Ni]O.O[Ni]O.[O-]C([O-])=O.[O-]C([O-])=O
InChI
1S/2CH2O3.5Ni.10H2O/c2*2-1(3)4;;;;;;;;;;;;;;;/h2*(H2,2,3,4);;;;;;10*1H2/q;;5*+2;;;;;;;;;;/p-10
InChI key
KWOUKTWMXYFKIW-UHFFFAOYSA-D
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.
Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.
Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.
Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service