Skip to Content
Merck
All Photos(3)

Documents

544183

Sigma-Aldrich

Nickel carbonate, basic hydrate

99.9% trace metals basis

Synonym(s):

Nickel carbonate hydroxide hydrate, Nickel(II) carbonate (basic) hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiCO3 · 2Ni(OH)2 · xH2O
CAS Number:
Molecular Weight:
304.12 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% (trace metals analysis)
99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: nickel

impurities

≤1500.0 ppm Trace Metal Analysis

application(s)

battery manufacturing

SMILES string

O.[Ni++].O[Ni]O.O[Ni]O.[O-]C([O-])=O

InChI

1S/CH2O3.3Ni.5H2O/c2-1(3)4;;;;;;;;/h(H2,2,3,4);;;;5*1H2/q;3*+2;;;;;/p-6

InChI key

CCFAFYHXJQHIRV-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

General description

Nickel carbonate, basic hydrate is a green powder that is water insoluble. It is the most important carbonate for industrial applications such as nickel electroplating, as well as an intermediate in the hydrometallurgical purification of nickel from its ores.

Application

Nickel carbonate, basic hydrate can be used as a precursor:
  • To synthesize spinel nickel cobaltite (NiCo2O4) nanoparticles for supercapacitor application.
  • To prepare nanocrystalline nickel disulfide via the solid-state reaction.
It can also be used as a catalyst to produce H2O2 on carbon cathode.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-assisted hydrothermal synthesis of spinel nickel cobaltite and application for supercapacitors
Chun-Chieh Tseng, et al.
Journal of the Taiwan Institute of Chemical Engineers, 44, 415-419 (2013)
H2O2 production on a carbon cathode loaded with a nickel carbonate catalyst and on an oxide photoanode without an external bias
Soichi Takasugi, et al.
Royal Society of Chemistry Advances, 11, 11224-11232 (2021)
Transition metal dichalcogenide materials: Solid-state reaction synthesis of nanocrystalline nickel disulfide
Gaojun An, et al.},
Materials Letters, 62, 2643-2646 (2008)

Articles

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service