Skip to Content
Merck
All Photos(1)

Documents

253731

Sigma-Aldrich

Phenethyl isothiocyanate

99%

Synonym(s):

2-Phenylethyl isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2NCS
CAS Number:
Molecular Weight:
163.24
Beilstein:
2084162
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.5888 (lit.)

bp

139-140 °C/11 mmHg (lit.)

density

1.094 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

S=C=NCCc1ccccc1

InChI

1S/C9H9NS/c11-8-10-7-6-9-4-2-1-3-5-9/h1-5H,6-7H2

InChI key

IZJDOKYDEWTZSO-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

Phenethyl isothiocyanate is a naturally occurring isothiocyanate in cruciferous vegetables. Phenethyl isothiocyanate is a chemopreventive agent. It inhibits cancer cell growth by arresting the cell cycle and inducing apoptosis in various human cancer cell models.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yating Wang et al.
Molecular medicine reports, 10(1), 543-549 (2014-05-03)
Phenethyl isothiocyanate (PEITC), a potential cancer chemopreventive constituent of cruciferous vegetables, including watercress, has been reported to inhibit cancer cell growth by arresting the cell cycle and inducing apoptosis in various human cancer cell models. However, the role of PEITC
Marilyn E Morris et al.
The AAPS journal, 16(4), 705-713 (2014-05-14)
Phenethyl isothiocyanate (PEITC)-a naturally occurring isothiocyanate in cruciferous vegetables-has been extensively studied as a chemopreventive agent in several preclinical species and in humans. Pharmacokinetic features of unchanged PEITC are (I) linear and first-order absorption, (II) high protein binding and capacity-limited
L D Huong et al.
Oral diseases, 18(5), 513-519 (2012-02-09)
β-Phenylethyl isothiocyanate (PEITC) has been demonstrated to fight many types of cancers through various molecular pathways. In this study, we focused on its effect on the induction of apoptosis to inhibit cell growth and molecular mechanism in oral cancer. 3-(4,5-dimethylthiazol-2-yl)-5-(2,4-disulfophenyl)-2-(4
Parul Gupta et al.
BMC medicine, 10, 80-80 (2012-07-25)
HER2 is an oncogene, expression of which leads to poor prognosis in 30% of breast cancer patients. Although trastuzumab is apparently an effective therapy against HER2-positive tumors, its systemic toxicity and resistance in the majority of patients restricts its applicability.
Wen-jing Wu et al.
Biochemical pharmacology, 85(4), 486-496 (2012-12-12)
Platinum (Pt)-based chemotherapy is an important regimen in the clinical treatment of cancer, but development of drug resistance presents a major challenge. One key mechanism involved in resistance to Pt drugs is the decrease of intracellular Pt due to the

Articles

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service