Skip to Content
Merck
All Photos(1)

Key Documents

36682

Supelco

Allyl isothiocyanate

PESTANAL®, analytical standard

Synonym(s):

AITC, Oil of mustard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2NCS
CAS Number:
Molecular Weight:
99.15
Beilstein:
773748
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

99.7% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.529 (lit.)

bp

150 °C (lit.)

mp

−80 °C (lit.)

density

1.013 g/mL at 25 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

C=CCN=C=S

InChI

1S/C4H5NS/c1-2-3-5-4-6/h2H,1,3H2

InChI key

ZOJBYZNEUISWFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Allyl isothiocyanate is a naturally occurring compound in plants which belongs to the class of cruciferae.[1] It can also serve as a promising candidate that can show anti-cancer activity in both cultured cells and animal models.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Naturally occurring constituent of cruciferous vegetables.

Packaging

bottled under inert gas

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antibacterial mechanism of allyl isothiocyanate
Lin C-M, et al.
Journal of Food Protection, 63, 727-734 (2000)
Allyl isothiocyanate as a cancer chemopreventive phytochemical
Zhang Y, et al.
Molecular Nutrition And Food Research, 54, 127-135 (2010)
Yuesheng Zhang
Molecular nutrition & food research, 54(1), 127-135 (2009-12-05)
Allyl isothiocyanate (AITC), which occurs in many common cruciferous vegetables, is widely and often frequently consumed by humans. Besides antimicrobial activity against a wide spectrum of pathogens, it showed anticancer activity in both cultured cancer cells and animal models, although
Enrico Morera et al.
Bioorganic & medicinal chemistry letters, 22(4), 1674-1677 (2012-01-20)
In order to explore the structural determinants for the TRPV1 and TRPA1 agonist properties of gingerols, a series of nineteen analogues (1b-5) of racemic [6]-gingerol (1a) was synthesized and tested on TRPV1 and TRPA1 channels. The exploration of the structure-activity
Meng Zhao et al.
Molecular pain, 8, 55-55 (2012-07-31)
Oxaliplatin, a platinum-based chemotherapeutic agent, causes an unusual acute peripheral neuropathy. Oxaliplatin-induced acute peripheral neuropathy appears in almost all patients rapidly after infusion, and is triggered or exacerbated by cold, while its mechanisms are poorly understood. In this study, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service