Skip to Content
Merck
All Photos(2)

Key Documents

244619

Sigma-Aldrich

Zinc selenide

powder, 10 μm, 99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSe
CAS Number:
Molecular Weight:
144.35
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

grade

deposition grade

Assay

99.99% trace metals basis

form

powder

particle size

10 μm

density

5.42 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Zn]=[Se]

InChI

1S/Se.Zn

InChI key

SBIBMFFZSBJNJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Shifu et al.
Journal of nanoscience and nanotechnology, 12(6), 4898-4904 (2012-08-22)
ZnS-CuS nanoparticles were prepared by a controlled coprecipitation method and ZnS-CuS-Ce2S3 nanoparticles were prepared by ball milling. The photocatalysts were characterized by scanning electron microscopy, X-ray powder diffraction, transmission electron microscopy, and terephthalic acid photoluminescence probing technique. The photocatalytic activity
Yunxing Du et al.
Journal of nanoscience and nanotechnology, 12(11), 8487-8493 (2013-02-21)
Nowadays, novel synthesis routes of nanoparticles are attracting a considerable attention of relative scientists. In this work, monodispersed spherical ZnS quantum dots (QDs) were synthesized by an oil-water interface method. The as-prepared products were characterized by X-ray diffraction (XRD), transmission
Jagdeep Kaur et al.
Journal of nanoscience and nanotechnology, 13(7), 4861-4871 (2013-08-02)
In the present work, Zinc sulphide (ZnS) nanoparticles (NPs) capped with 2-Mercaptoethanol (2 ME) are synthesized via chemical precipitation method. Structural and morphological studies have been done using X Ray Diffraction (XRD) and Transmission Electron Microscopy (TEM) techniques. Band gap
Zhaoxia Cai et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 97, 909-914 (2012-08-21)
Water-soluble and high luminescent l-glutathione-capped-ZnSe quantum dots (QDs) were applied for ultrasensitive Pb(II) detection. l-glutathione-capped-ZnSe QDs were synthesized by a facile colloid aqueous phase route. It is characterized by transmission electron microscopy, X-ray powder diffraction spectrometry, Fourier transform infrared spectrometry
Arvind P Ravikumar et al.
Optics express, 20(20), 22391-22397 (2012-10-06)
We report the design, fabrication and characterization of a II-VI Zn(0.51)Cd(0.49)Se / Zn<sub>0.45(Cd)0.42(Mg)(0.13)Se-based quantum well infrared photodetector (QWIP) with a bound to quasi-bound transition centered at 8.7 µm. The good growth quality of the epitaxial layers was verified by x-ray

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service