Skip to Content
Merck
All Photos(2)

Key Documents

230014

Sigma-Aldrich

Zinc iodide

≥99.99% trace metals basis

Synonym(s):

Diiodozinc, Zinc diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnI2
CAS Number:
Molecular Weight:
319.20
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.99% trace metals basis
form:
crystalline

Quality Level

Assay

≥99.99% trace metals basis

form

crystalline

reaction suitability

reagent type: catalyst
core: zinc

impurities

≤100.0 ppm Trace Metal Analysis

mp

445 °C (lit.)

density

4.74 g/mL at 25 °C (lit.)

SMILES string

I[Zn]I

InChI

1S/2HI.Zn/h2*1H;/q;;+2/p-2

InChI key

UAYWVJHJZHQCIE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Zinc iodide is a faint yellow solid that absorbs water readily. It is synthesized using metallic zinc with an aqueous iodine solution. It is used as a Lewis acid for organic conversion.

Application

Zinc iodide can be used as:
  • An electrolyte in Zinc-iodine redox flow batteries.
  • An additive to prepare air-stable cationic lead perovskite films. The addition of ZnI2 enhances the film stability and photoluminescence without affecting the bandgap.
  • As a precursor to prepare efficient, eco-friendly, and high color purity inkjet-printed blue InP/ZnS/ZnS quantum dot light-emitting diodes.
  • A catalyst in promoting various chemical transformations, for example, in the direct synthesis of 2,3-Dihydroisoxazoles via a [3+2] cycloaddition reaction of the nitrones and terminal alkynes.
  • An activator/initiator in living cationic polymerization reactions.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Inkjet-printed blue InP/ZnS/ZnS quantum dot light-emitting diodes
Zhiwen Yang, et al.
Chemical Engineering Journal, 450, 138413-138413 (2022)
Influence of type of zinc salts on photoinitiated living cationic polymerization of vinyl ethers
Muhammet U. Kahveci, et al.
Polymer, 54, 4798-4801 (2013)
Air-Stable and Oriented Mixed Lead Halide Perovskite (FA/MA) by the One-Step Deposition Method Using Zinc Iodide and an Alkylammonium Additive
Loreta A. Muscarella, et al.},
ACS Applied Materials & Interfaces, 11, 17555-17562 (2019)
Zinc Iodide-Mediated Direct Synthesis of 2,3-Dihydroisoxazoles from Alkynes and Nitrones
Zu-Feng Xiao, et al.
advanced synthesis and catalysis, 358, 1859-1863 (2016)
Metal?Organic Frameworks as Highly Active Electrocatalysts for High-Energy Density, Aqueous Zinc-Polyiodide Redox Flow Batteries
Bin Li, et al.
Nano Letters, 16, 4335-4340 (2016)

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service