Skip to Content
Merck
All Photos(1)

Documents

96483

Sigma-Aldrich

Zinc iodide

purum p.a., ≥98.0% (AT)

Synonym(s):

Diiodozinc, Zinc diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnI2
CAS Number:
Molecular Weight:
319.20
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

purum p.a.

Assay

≥98.0% (AT)

form

powder or crystals

mp

445 °C (lit.)

density

4.74 g/mL at 25 °C (lit.)

anion traces

sulfate (SO42-): ≤500 mg/kg

cation traces

Ca: ≤500 mg/kg
Cd: ≤200 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤100 mg/kg
K: ≤500 mg/kg
Na: ≤500 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg

SMILES string

I[Zn]I

InChI

1S/2HI.Zn/h2*1H;/q;;+2/p-2

InChI key

UAYWVJHJZHQCIE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Zinc iodide is a white crystalline Lewis acid often used as a starting material for synthesizing other zinc compounds, as a Lewis acid catalyst in organic reactions, and as an electrolyte additive.

Application

Zinc iodide can be used:
  • As a catalyst for one-pot synthesis of aminoindolizines via sequential A3 coupling/cycloisomerization.
  • As a catalyst for one-pot synthesis of quinoxalines.
  • As a precursor to synthesize highly luminescent quantum dots.
  • As a redox electrolyte in hybrid energy storage systems.
  • As a dopant to prepare highly flexible PVA composite films. The addition of zinc iodide enhances the optoelectronic properties of polymer composite.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient one-pot synthesis of quinoxalines in the presence of zinc iodide as catalyst
J N Sangshetti, et al.
Russian Journal of Organic chemistry, 45, 1116-1118 (2009)
The effect of zinc iodide on the physicochemical properties of highly flexible transparent poly (vinyl alcohol) based polymeric composite films: opto-electrical performance
Bouzidi A , et al.
Journal of Materials Science: Materials in Electronics, 30, 11799-11806 (2019)
Nanoconfinement of redox reactions enables rapid zinc iodide energy storage with high efficiency
Juhan Lee, et al.
Journal of Materials Chemistry, 5, 12520-12527 (2017)
Zinc iodide: a mild and efficient catalyst for one-pot synthesis of aminoindolizines via sequential A 3 coupling/cycloisomerization
Subhajit Mishra, et al.
Royal Society of Chemistry Advances, 4, 6672-6676 (2014)
Large-Scale Synthesis of Highly Luminescent InP@ZnS Quantum Dots Using Elemental Phosphorus Precursor
Eunbyul Bang, et al.
Chemistry of Materials, 29, 4236-4243 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service