Skip to Content
Merck
All Photos(1)

Key Documents

220027

Sigma-Aldrich

3-Indolepropionic acid

ReagentPlus®, 99%

Synonym(s):

3-(3-Indolyl)propanoic acid, IPA, Indole-3-propionic acid, NSC 3252, NSC 47831

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO2
CAS Number:
Molecular Weight:
189.21
Beilstein:
147733
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

product line

ReagentPlus®

Assay

99%

form

powder

mp

134-135 °C (lit.)

solubility

ethanol: soluble 50 mg/mL, clear, yellow to orange

functional group

carboxylic acid

SMILES string

OC(=O)CCc1c[nH]c2ccccc12

InChI

1S/C11H11NO2/c13-11(14)6-5-8-7-12-10-4-2-1-3-9(8)10/h1-4,7,12H,5-6H2,(H,13,14)

InChI key

GOLXRNDWAUTYKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Indolepropionic acid is an effective inhibitor of aggregation of misfolded β-amyloid protein (Abeta)[1]. Three-component one-pot procedure has been reported to assemble 3-indolepropionic acids[2].

Application

Reactant for preparation of:
  • Fluorescent analogues of strigolactones
  • Anti-tumor agents
  • Melanocortin receptors ligands
  • Immunosuppressive agents
  • Iinhibitors of hepatitis C virus
  • Histamine H4 receptor agonists
  • NR2B/NMDA receptor antagonists
  • CB1 Antagonist for the treatment of obesity
  • Antibacterial agents
  • Inhibitor of TGF-β receptor binding

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mauro F A Adamo et al.
Organic letters, 9(2), 303-305 (2007-01-16)
A three-component one-pot procedure (3-MC) was developed to assemble 3-indolepropionic acids from commercially available materials. This new methodology affords the title compounds in high yields and without the use of chromatography. [reaction: see text].
Xun Cheng et al.
Analytical chemistry, 77(21), 7012-7015 (2005-11-01)
Alzheimer's disease is the most common cause of the loss of cognitive function among the elderly, and the aggregation and deposition of misfolded beta-amyloid protein (Abeta) contribute to this progressive central nervous system decline. Therefore, compounds that inhibit or even
F Mandelbaum-Shavit et al.
Antimicrobial agents and chemotherapy, 35(12), 2526-2530 (1991-12-01)
Indole-3-propionic acid (IPA), a phytohormone derivative, is a potent inhibitor of growth of Legionella pneumophila cultivated extracellularly in a chemically defined hypotonic medium and intracellularly in human monocytes. The inhibitory activity turns into bactericidal activity with increasing concentrations. The susceptibility
Qiaomei Lu et al.
Rapid communications in mass spectrometry : RCM, 22(16), 2565-2572 (2008-07-26)
Auxin is an important phylohormone, which regulates specific physiological responses such as division, elongation and differentiation of cells. A new method using liquid chromatography/electrospray ionization ion trap mass spectrometry (LC/ESI-ITMS) has been developed for identification and quantitation of four auxins.
Paul E Bendheim et al.
Journal of molecular neuroscience : MN, 19(1-2), 213-217 (2002-09-06)
The accumulation of amyloid-beta and concomitant oxidative stress are major pathogenic events in Alzheimer's disease. Indole-3-propionic acid (IPA, OXIGON) is a potent anti-oxidant devoid of pro-oxidant activity. IPA has been demonstrated to be an inhibitor of beta-amyloid fibril formation and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service