Skip to Content
Merck
All Photos(1)

Key Documents

I1625

Sigma-Aldrich

3-Indoleacrylic acid

powder

Synonym(s):

3-(3-Indolyl)acrylic acid, IAA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9NO2
CAS Number:
Molecular Weight:
187.19
Beilstein:
6317
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.52

grade

Molecular Biology
for molecular biology

Assay

≥98.0% (HPLC)
≥98.0% (TLC)

form

powder

mol wt

187.19  g/mol

storage temp.

room temp

SMILES string

OC(=O)\C=C\c1c[nH]c2ccccc12

InChI

1S/C11H9NO2/c13-11(14)6-5-8-7-12-10-4-2-1-3-9(8)10/h1-7,12H,(H,13,14)/b6-5+

InChI key

PLVPPLCLBIEYEA-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Indoleacrylic acid (IAA) is a chemical used for the induction of gene transcription. This expression system requires that the gene of interest is cloned in plasmids containing the trpE promoter.

Application

Suitable for induction of gene transcription in systems controlled by the trpE promoter.

Biochem/physiol Actions

3-Indoleacrylic acid helps to block the mycelial growth of Neurospora crassa. As a result, the cells accumulate indoleglycerol phosphate, which influences the rate of tryptophan synthetase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of tryptophan synthetase by indoleacrylic acid
Matchett W H
Journal of Bacteriology, 110(1), 146-154 (1972)
D N Wilson et al.
RNA (New York, N.Y.), 6(12), 1704-1713 (2001-01-06)
Replacing a cassette of 31 residues from Escherichia coli release factor 1 with the equivalent residues in release factor 2 gave a protein active in codon-specific binding to the ribosome but inactive in peptidyl-tRNA hydrolysis. Such a phenotype is also
Patricia Casino et al.
Biochemistry, 46(26), 7728-7739 (2007-06-15)
Substrate channeling in the tryptophan synthase bienzyme complex from Salmonella typhimurium is regulated by allosteric interactions triggered by binding of ligand to the alpha-site and covalent reaction at the beta-site. These interactions switch the enzyme between low-activity forms with open
T Mohammad et al.
Photochemistry and photobiology, 59(2), 189-196 (1994-02-01)
Naked, infectious single-stranded (ss) and double-stranded (ds) DNA from phages S13 and G4 were irradiated with 308 nm UV radiation in the absence and presence of several photobiologically active compounds: E- and Z-urocanic acid (E- and Z-UA), their methyl esters
J G Bell et al.
Prostaglandins, leukotrienes, and essential fatty acids, 63(1-2), 21-25 (2000-09-06)
The fatty acid compositions of red blood cell (RBC) phospholipids from a patient with autistic spectrum disorder (ASD) had reduced percentages of highly unsaturated fatty acids (HUFA) compared to control samples. The percentage of HUFA in the RBC from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service