Skip to Content
Merck

57400

Indole-3-propionic acid

≥99.0% (T)

Synonym(s):

3-(3-Indolyl)propanoic acid, 3-(3-Indolyl)propionic acid, IPA, NSC 3252, NSC 47831

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H11NO2
CAS Number:
Molecular Weight:
189.21
UNSPSC Code:
12352106
NACRES:
NA.77
PubChem Substance ID:
EC Number:
212-600-1
Beilstein/REAXYS Number:
147733
MDL number:
Assay:
≥99.0% (T)
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99.0% (T)

form

solid

ign. residue

≤0.1%

mp

~133 °C

SMILES string

OC(=O)CCc1c[nH]c2ccccc12

InChI

1S/C11H11NO2/c13-11(14)6-5-8-7-12-10-4-2-1-3-9(8)10/h1-4,7,12H,5-6H2,(H,13,14)

InChI key

GOLXRNDWAUTYKT-UHFFFAOYSA-N

Application


  • Elucidating the Interaction of Indole-3-Propionic Acid and Calf Thymus DNA: Multispectroscopic and Computational Modeling Approaches.: This study investigates the binding interaction between indole-3-propionic acid and calf thymus DNA, employing multispectroscopic techniques and computational modeling. The insights gained could lead to a deeper understanding of the molecular mechanisms underpinning the protective effects of this compound on DNA integrity, critical for developing novel therapeutic strategies in biochemistry (Wei Y et al., 2024).

  • The Tryptophan Metabolite Indole-3-Propionic Acid Raises Kynurenic Acid Levels in the Rat Brain In Vivo.: This research demonstrates how indole-3-propionic acid, a metabolite of tryptophan, can increase levels of kynurenic acid in the brain, offering potential benefits in neuroprotective strategies and the study of psychiatric disorders. Such findings highlight significant implications for mental health research and therapeutic applications (Sathyasaikumar KV et al., 2024).

  • Serum-Derived Bovine Immunoglobulin Promotes Barrier Integrity and Lowers Inflammation for 24 Human Adults Ex Vivo.: This study explores how indole-3-propionic acid impacts inflammatory responses and gut barrier integrity, providing valuable data for nutritional science and clinical applications aimed at reducing gastrointestinal disorders and enhancing overall gut health (Van den Abbeele P et al., 2024).

  • The Effect of Type 2 Resistant Starch and Indole-3-Propionic Acid on Ameliorating High-Fat-Diet-Induced Hepatic Steatosis and Gut Dysbiosis.: This publication discusses the beneficial effects of indole-3-propionic acid in conjunction with type 2 resistant starch on reducing hepatic steatosis and correcting gut dysbiosis caused by high-fat diets, pointing to potential dietary interventions for managing obesity and associated metabolic disorders (Yang M et al., 2024).

  • Gut microbiome and cardiometabolic comorbidities in people living with HIV.: This review highlights the role of indole-3-propionic acid in modulating the gut microbiome and its potential effects on cardiometabolic health in HIV-positive individuals. The findings suggest a promising avenue for improving the quality of life and health outcomes in this population through microbiome-based interventions (Trøseid M et al., 2024).

Biochem/physiol Actions

Studied as an adjunct to improve perfusion after liver transplant.
Was studied as an adjunct to improve perfusion after liver transplant.


Still not finding the right product?

Explore all of our products under Indole-3-propionic acid


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library