Skip to Content
Merck
All Photos(1)

Key Documents

I5508

Sigma-Aldrich

DL-Indole-3-lactic acid

99%

Synonym(s):

β-(3-Indolyl)lactic acid, 2-Hydroxy-3-(3-indolyl)propanoic acid, 3-(3-Indolyl)-2-hydroxypropanoic acid, 3-Indolyllactic acid, Indolelactic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO3
CAS Number:
Molecular Weight:
205.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

145-146 °C (lit.)

SMILES string

OC(Cc1c[nH]c2ccccc12)C(O)=O

InChI

1S/C11H11NO3/c13-10(11(14)15)5-7-6-12-9-4-2-1-3-8(7)9/h1-4,6,10,12-13H,5H2,(H,14,15)

InChI key

XGILAAMKEQUXLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DL-Indole-3-lactic acid can serve as a building block in the synthesis of various molecules, like Indole-3-acetic acid (IAA)

Application

Reactant for preparation of:
  • Antibacterial agents
  • Dietary sweetener

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of urinary 5-hydroxyindole-3-acetic acid using solid-phase extraction and reversed-phase high-performance liquid chromatography with electrochemical detection.
P P Chou et al.
Journal of chromatography, 341(1), 167-171 (1985-05-31)
Soumen K Manna et al.
Journal of proteome research, 9(8), 4176-4188 (2010-06-15)
Alcohol-induced liver disease (ALD) is a leading cause of nonaccident-related deaths in the United States. Although liver damage caused by ALD is reversible when discovered at the earlier stages, current risk assessment tools are relatively nonspecific. Identification of an early
Francisco A Moreno et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 20(1), 18-24 (2009-11-10)
The purpose of this study was to examine the differential effects of acute tryptophan (TRP) depletion vs. sham condition on plasma, cerebrospinal fluid (CSF) biochemical parameters, and mood in the following three subject groups: (1) nine antidepressant-free individuals with remitted
S Krásenský et al.
Electrophoresis, 16(6), 968-973 (1995-06-01)
A new approach is described for highly sensitive chiral analyses by capillary zone electrophoresis, based on using an on-line combination of two capillaries filled with either chiral selective or achiral background electrolytes (BGE). Thus, the BGEs are selected in such
X Liu et al.
The Journal of biological chemistry, 269(1), 668-675 (1994-01-07)
The hormone, auxin, plays an important role in the differentiation and growth of plant cells. A number of auxin-responsive genes have been characterized but until now minimal auxin-responsive cis-elements within these promoter regions have not been identified. Here we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service