Skip to Content
Merck
All Photos(2)

Key Documents

205494

Sigma-Aldrich

Dibutyltin dichloride

96%

Synonym(s):

Dibutyldichlorotin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)2SnCl2
CAS Number:
Molecular Weight:
303.84
Beilstein:
3535484
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

bp

135 °C/10 mmHg (lit.)

mp

37-40 °C (lit.)

SMILES string

CCCC[Sn](Cl)(Cl)CCCC

InChI

1S/2C4H9.2ClH.Sn/c2*1-3-4-2;;;/h2*1,3-4H2,2H3;2*1H;/q;;;;+2/p-2

InChI key

RJGHQTVXGKYATR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

  • Effects of dibutyltin compounds on PPARγ/RXRα activity: Studies the impact of various dibutyltin compounds, including dibutyltin dichloride, on adipogenesis and inflammation in mammalian cells, emphasizing their effects on the PPARγ/RXRα pathway (Milton et al., 2017).

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Muta. 2 - Repr. 1B - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 Oral - STOT SE 1 Oral

Target Organs

thymus

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Gisela Sparmann et al.
Cell research, 20(3), 288-298 (2010-01-27)
Origin and fate of pancreatic stellate cells (PSCs) before, during and after pancreatic injury are a matter of debate. The crucial role of PSCs in the pathogenesis of pancreatic fibrosis is generally accepted. However, the turnover of the cells remains
Louis P Vera-Portocarrero et al.
Gastroenterology, 135(4), 1369-1378 (2008-08-13)
Sumatriptan is used specifically to relieve headache pain. The possible efficacy of sumatriptan was investigated in 2 models of visceral pain. Pancreatic inflammation was induced by intravenous injection of dibutyltin dichloride. Noninflammatory irritable bowel syndrome was induced by intracolonic instillation
Kent J Fanning et al.
Antimicrobial agents and chemotherapy, 55(6), 3008-3011 (2011-03-16)
This work sought to define how pancreatitis affected antibiotic distribution in a perfused rat pancreas model. The distribution kinetics of four antibiotics were examined in control animals and animals with pancreatitis. Meropenem and piperacillin distributed into the extracellular space, and
Guoqing Shi et al.
Environmental health perspectives, 117(3), 379-386 (2009-04-02)
Because of the vital importance of the proteasome pathway, chemicals affecting proteasome activity could disrupt essential cellular processes. Although the toxicity of organotins to both invertebrates and vertebrates is well known, the essential cellular target of organotins has not been
Toyoma Kaku et al.
Pancreas, 34(3), 299-309 (2007-04-07)
Chronic pancreatitis consists of excessive leukocyte infiltration and fibrosis. IS-741 has been reported to be an antiinflammatory drug through an inhibitory action on cell adhesion. In this study, we investigated whether IS-741 could inhibit the progression of pancreatic fibrosis through

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service