Skip to Content
Merck
All Photos(1)

Key Documents

46105

Supelco

Irgarol®

PESTANAL®, analytical standard

Synonym(s):

2-(tert-Butylamino)-4-(cyclopropylamino)-6-(methylthio)-s-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19N5S
CAS Number:
Molecular Weight:
253.37
Beilstein:
792218
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

CSc1nc(NC2CC2)nc(NC(C)(C)C)n1

InChI

1S/C11H19N5S/c1-11(2,3)16-9-13-8(12-7-5-6-7)14-10(15-9)17-4/h7H,5-6H2,1-4H3,(H2,12,13,14,15,16)

InChI key

HDHLIWCXDDZUFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Irgarol is a registered trademark of Ciba Specialty Chemicals Corp.
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hassan Rashid Ali et al.
Marine pollution bulletin, 70(1-2), 253-257 (2013-03-16)
Emerging booster biocides contamination raises particular attention in the marine ecosystem health. This study provides the baseline data on the occurrence of Irgarol-1051 (2-methylthio-4-tert-butylamino-6-cyclopropylamiono-s-triazine) in the selected coastal water around Malaysia. The maximum detected concentration of Irgarol was 2021 ng/L
Niki C Maragou et al.
Journal of the American Society for Mass Spectrometry, 22(10), 1826-1838 (2011-09-29)
A systematic and detailed optimization strategy for the development of atmospheric pressure ionization (API) LC-MS/MS methods for the determination of Irgarol 1051, Diuron, and their degradation products (M1, DCPMU, DCPU, and DCA) in water, sediment, and mussel is described. Experimental
Vivien W W Bao et al.
Marine pollution bulletin, 62(5), 1147-1151 (2011-03-23)
Since 1990s, various booster biocides have been increasingly used as substitutes of organotins. However, knowledge about their toxicities on tropical/sub-tropical marine species is significantly lacking. This study comprehensively investigated the acute toxicities of copper, tributyltin (TBT), and five commonly used
Hiroya Harino et al.
Archives of environmental contamination and toxicology, 63(1), 13-21 (2012-05-10)
Coastal marine environments are considered to be the most sensitive areas for the accumulation of organotin (OT) compounds and other emerging new pollutants, such as perfluoroalkyl compounds. Contamination by these compounds is a matter of great concern due to their
Melissa Faria et al.
The Science of the total environment, 408(12), 2452-2458 (2010-03-30)
This study evaluated the impact of biocides (tributyltin, chlorthalonil and Irgarol 1051) and of pollutants (copper, inorganic and methyl mercury and 4-nonylphenol) occurring in Ebro River (NE Spain) on early developmental stages of native Spanish freshwater and invasive zebra mussels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service