Skip to Content
Merck
All Photos(3)

Documents

183083

Sigma-Aldrich

Dibutyltin(IV) oxide

98%

Synonym(s):

Dibutyloxotin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)2SnO
CAS Number:
Molecular Weight:
248.94
Beilstein:
4126243
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

autoignition temp.

534 °F

mp

≥300 °C (lit.)

SMILES string

CCCC[Sn](=O)CCCC

InChI

1S/2C4H9.O.Sn/c2*1-3-4-2;;/h2*1,3-4H2,2H3;;

InChI key

JGFBRKRYDCGYKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibutyltin(IV) oxide is an organotin compound that is commonly used as a Lewis acid catalyst in organic synthesis. For instance, it can be used for the formation of carbon-carbon bonds through coupling reactions and in the preparation of esters from carboxylic acids and alcohols. It also finds use in allylation reactions, cycloaddition reactions, and ring-opening polymerizations.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 - STOT SE 1

Target Organs

thymus

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Ishido et al.
Nucleic acids symposium series, (8)(8), s7-s8 (1980-01-01)
For partial phenylcarbamoylation of the hydroxy-groups of ribonucleosides, dibutyltin oxide--phenyl isocyanate system was found to be surperior to the bis(tributyltin) oxide--phenyl isocyanate system from the standpoint of reaction procedures including isolation of the products; the reaction was proved to occur
K Takeo et al.
Carbohydrate research, 278(2), 301-313 (1995-12-20)
Dibutyltin oxide-mediated regioselective chloroacetylation of methyl 1-thio-beta-xylobioside, followed by treatment of the product with 4-methylbenzoyl chloride-pyridine, gave methyl 4-O-chloroacetyl-2,3-di-O-(4-methylbenzoyl)-beta-D-xylopyranosyl-(1-->4) -2.3-di - O-(4-methylbenzoyl)-1-thio-beta-D-xylopyranoside (18) in 70% yield. Coupling of 18 with benzyl alcohol afforded the disaccharide benzyl beta-glycoside, which was O-dechloroacetylated
K Takeo et al.
Carbohydrate research, 277(2), 231-244 (1995-11-22)
2- and 4-Nitrophenyl beta-D-xylopyranosides (4 and 5) were transformed, via dibutyltin oxidemediated acylation, into the corresponding 2,3-di-O-benzoyl derivatives 11 and 15. Xylobiose and xylotriose were easily isolated by charcoal column chromatography from a commercially available material and converted into the
Hong-Min Liu et al.
Carbohydrate research, 337(19), 1763-1767 (2002-11-09)
A mild and efficient neutral method for the cleavage of O-acetyl groups with dibutyltin oxide has been developed. This method is especially useful in the synthesis of glycosides containing base- or acid-sensitive multifunctional groups.
K Kiyoshima et al.
Chemical & pharmaceutical bulletin, 37(4), 861-865 (1989-04-01)
4''-O-Acyl-, 4''-O-alkyl- and 4''-deoxy-tylosin derivatives were synthesized using 2'-O-acetyl-3'',4''-O-(dibutyl-stannio)tylosin as a synthetic intermediate. The in vitro biological evaluation showed that the new derivatives were active against macrolide-resistant clinical isolates of bacteria and mycoplasmas, and that they were resistant to hepatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service