Skip to Content
Merck
All Photos(1)

Key Documents

T50202

Sigma-Aldrich

Tributyltin chloride

96%

Synonym(s):

TBTC, Tributylchlorotin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3SnCl
CAS Number:
Molecular Weight:
325.51
Beilstein:
3535715
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

<0.01 mmHg ( 20 °C)

Quality Level

Assay

96%

refractive index

n20/D 1.492 (lit.)

bp

171-173 °C/25 mmHg (lit.)

density

1.2 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](Cl)(CCCC)CCCC

InChI

1S/3C4H9.ClH.Sn/c3*1-3-4-2;;/h3*1,3-4H2,2H3;1H;/q;;;;+1/p-1

InChI key

GCTFWCDSFPMHHS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Tributyltin chloride (Bu3SnCl or tri-n-butylstannane) is an organotin compound that can be used as a reagent for:
  • The synthesis of fluorinated stannanes by reacting with fluorinated alkyl, aryl or vinyl halides using Zn or Cd catalyst via Barbier-type reaction.
  • The preparation of γ-hydroxyvinylstannanes, and indol-2-yltributylstannane.
  • The conversion of organozirconiums to organostannanes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorinated stannanes: Part 2. The stereospecific synthesis of fluorinated stannanes via a Barbier-type reaction between fluorinated halides and tributyltin chloride mediated by zinc or cadmium
Burton DJ and Jairaj V
Journal of Fluorine Chemistry, 126(5), 797-801 (2005)
Rita Jordão et al.
Environmental health perspectives, 123(8), 813-819 (2015-03-25)
The analysis of obesogenic effects in invertebrates is limited by our poor knowledge of the regulatory pathways of lipid metabolism. Recent data from the crustacean Daphnia magna points to three signaling hormonal pathways related to the molting and reproductive cycles
Transmetalation from zirconium to tin: A facile preparation of organostannanes from organozirconiums
Kim S and Kim KH
Tetrahedron Letters, 36(21), 3725-3728 (1995)
A Route to the Preparation of γ -Hydroxyvinylstannanes
Lautens M and Huboux AH
Tetrahedron Letters, 31(22), 3105-3108 (1990)
Julia F P de Araújo et al.
Toxicology letters, 295, 99-114 (2018-06-18)
Tributyltin chloride (TBT) is an obesogen associated with various metabolic and reproductive dysfunctions after in utero exposure. However, few studies have evaluated TBT's obesogenic effect on adult ovaries. In this study, we assessed whether TBT's obesogenic effects resulted in adult

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service