Merck
Catalytic antioxidants: regenerable tellurium analogues of vitamin E.
Organic letters (2013-11-28)
Vijay P Singh, Jia-fei Poon, Lars Engman
ABSTRACT
In an effort to improve the chain-breaking capacity of the natural antioxidants, an octyltelluro group was introduced next to the phenolic moiety in β- and δ-tocopherol. The new vitamin E analogues quenched peroxyl radicals more efficiently than α-tocopherol and were readily regenerable by aqueous N-acetylcysteine in a simple membrane model composed of a stirring chlorobenzene/water two-phase system. The novel tocopherol analogues could also mimic the action of the glutathione peroxidase enzymes.
Sigma-Aldrich
Tellurium, pieces, 99.999% trace metals basis
Sigma-Aldrich
Tellurium, powder, −200 mesh, 99.8% trace metals basis
Sigma-Aldrich
Tellurium, shot, ≤2 mm, 99.999% trace metals basis
Sigma-Aldrich
Tellurium, powder, −30 mesh, 99.997% trace metals basis
Sigma-Aldrich
Tellurium, granular, −5-+50 mesh, 99.99% trace metals basis