Skip to Content
Merck
All Photos(3)

Key Documents

205338

Sigma-Aldrich

Tellurium tetrachloride

99%

Synonym(s):

Tellurium chloride, Tellurium(IV) chloride, Tetrachlorotellurium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TeCl4
CAS Number:
Molecular Weight:
269.41
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder, crystals or chunks

Quality Level

Assay

99%

form

powder, crystals or chunks

bp

380 °C (lit.)

mp

224 °C (lit.)

density

3.26 g/mL at 25 °C (lit.)

SMILES string

Cl[Te](Cl)(Cl)Cl

InChI

1S/Cl4Te/c1-5(2,3)4

InChI key

SWLJJEFSPJCUBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Atomic number of base material: 52 Tellurium

Application

  • Highly efficient regioselective synthesis of organotellurium compounds: Explores the reactions of Tellurium Tetrachloride with 1-alkenes, highlighting its utility in organic synthesis (VA Potapov et al., 2017).
  • Cyclization Reactions: TeCl4 acts as a bis-electrophile in double-electrophilic cyclization reactions, demonstrating its versatility in organic transformations (N Korol, M Slivka, 2018).
  • Regio-and stereoselective addition of tellurium tetrachloride: Investigates the addition of TeCl4 to methyl propargyl ether, emphasizing the stereochemistry of the addition process (MV Musalova et al., 2017).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Puneet Vij et al.
Environmental toxicology and pharmacology, 34(3), 768-782 (2012-10-17)
Diphenyl ditelluride (DPDT) and tellurium tetrachloride (TeCl(4)) were evaluated for toxicity in transformed (HT-29, Caco-2) and non-transformed colon cells (CCD-18Co). Significant decreases in viability were observed with DPDT exposure in HT-29 (62.5-1000 μM), Caco-2 (31.25-1000 μM) and CCD-18Co cells (500-1000
J F Van Vleet et al.
American journal of veterinary research, 43(11), 2000-2009 (1982-11-01)
Seventy newly hatched ducklings were fed a commercial ration with 500 mg of added Te (as tetrachloride)/kg of feed for up to 28 days. Ducklings were euthanatized at day 14, 21, and 28, the hearts were studied by gross, microscopic
P Mareeswari et al.
Enzyme and microbial technology, 95, 225-229 (2016-11-22)
We report an efficient method to biosynthesize biocompatible cadmium telluride and cadmium sulphide quantum dots from the fungus Rhizopus stolonifer. The suspension of the quantum dots exhibited purple and greenish-blue luminescence respectively upon UV light illumination. Photoluminescence spectroscopy, X-ray diffraction
Shalini Roy et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 49(10), 2564-2574 (2011-07-12)
Tellurium tetrachloride (TeCl(4)) and diphenyl ditelluride (DPDT) cytotoxicity, was investigated in rat astrocytes. Concentrations of 0.24-250μM (24h) were tested for viability using MTT(3-[4,5-dimethylthiazol-2-yl]-2,5-diphenyl tetrazolium bromide) and trypan blue exclusion. MTT showed significant decreases at all concentrations tested for both compounds.
Maria V Musalova et al.
Molecules (Basel, Switzerland), 17(5), 5770-5779 (2012-05-17)
The reaction of tellurium tetrachloride with acetylene proceeds in a stereospecific anti-addition manner to afford the novel products E-2-chlorovinyltellurium trichloride and E,E-bis(2-chlorovinyl)tellurium dichloride. Reaction conditions for the selective preparation of each of these products were found. The latter was obtained

Articles

Thin film photovoltaic devices have become increasingly important in efficiently harnessing solar energy to meet consumer demand.

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service