Merck
Teratogenic vulnerability of Wistar rats to diphenyl ditelluride.
Toxicology (2004-12-15)
E C Stangherlin, A M Favero, G Zeni, J B T Rocha, C W Nogueira
ABSTRACT
The effect of single maternal subcutaneous (s.c.) injection of 0.12 mg/kg diphenyl ditelluride, (PhTe)2, diluted in canola oil at days 6, 10 or 17 of gestation were evaluated in Wistar rats. The reduction of body weight gain was statistically significant at GD9, for the dams that received (PhTe)2, at GD6; at GD13, for the dams that received (PhTe)2, at GD10, and at GD20, for the dams that received (PhTe)2, at GD17, when compared to respective control groups. External and internal fetal soft tissues examination was performed on day 20 of gestation. Single maternal injection at day 10 of gestation resulted in appearance of malformation in fore- and hind-limbs, absent or short tail, subcutaneous blood clots, exophthalmia, hydrocephalus and absence of the cranial bone and cutaneous tissue in fetuses on day 20 of gestation. Besides, (PhTe)2 reduced fetal body and cerebral weight, kidney length, measurements of body dimension and provoked 73% of fetal mortality. Subcutaneous administration of (PhTe)2 on day 17 of gestation was associated with 94% mortality, hydrocephalus and edema. Histological evaluations of fetal brain demonstrated displaced brain tissue with absence of the cranial bone and cutaneous tissue when diphenyl ditelluride was administered in GD10. Histological evaluation of fetal head exposed at GD17 revealed a decrease of the brain volume with consequent dilation of the lateral ventricles and the adjacent tissues were thinner than that of control group tissues. No fetal changes were observed after administration of (PhTe)2 at day 6 of gestation. Thus, (PhTe)2 can be teratogenic to rat fetuses and toxic for dams. The late fetal stages of rat prenatal development appeared uniquely sensitive to organic tellurium exposure.
Sigma-Aldrich
Tellurium, pieces, 99.999% trace metals basis
Sigma-Aldrich
Tellurium, shot, ≤2 mm, 99.999% trace metals basis
Sigma-Aldrich
Tellurium, powder, −200 mesh, 99.8% trace metals basis
Sigma-Aldrich
Tellurium, powder, −30 mesh, 99.997% trace metals basis
Sigma-Aldrich
Tellurium, granular, −5-+50 mesh, 99.99% trace metals basis