Skip to Content
Merck
All Photos(4)

Documents

176141

Sigma-Aldrich

4-Hydroxy-TEMPO

95%

Synonym(s):

4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxyl, TEMPOL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18NO2
CAS Number:
Molecular Weight:
172.24
Beilstein:
1422990
EC Number:
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

composition

Carbon content, 60.6-65.0%
Nitrogen content, 7.8-8.4%

mp

69-71 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)CC(O)CC(C)(C)N1[O]

InChI

1S/C9H18NO2/c1-8(2)5-7(11)6-9(3,4)10(8)12/h7,11H,5-6H2,1-4H3

InChI key

UZFMOKQJFYMBGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Hydroxy-TEMPO is a 4-substituted 2,2,6,6-tetramethylpiperidyl-1-oxy (TEMPO) derivative. It is a low-molecular weight compound and has been proposed as superoxide dismutase mimic.

Application

4-Hydroxy-TEMPO (HO-TEMPO) may be employed as catalyst for the oxidation of alcohols to the corresponding aldehydes. It may be employed for the preparation of TEMPO based polymer catalyst systems, which are useful for the Anelli oxidation of various primary alcohols. It may be used as starting reagent in the synthesis of 4-(2,2,6,6-tetramethyl-1-oxyl-4-piperidoxyl)butyl bromide.
A fluorous-tagged TEMPO derivative was prepared from the derived TEMPO propargylic ether and subsequent "click" reaction with a fluorous azide. This TEMPO derivative proved to be a highly effective catalyst in the oxidation of alcohols with bleach.
Spin label for studying biological compounds and polymers.
Spin label reagent used in the chatacterization of antagonist and agonist binding sites of NK1 receptor. Also used to measure reactive oxygen generation in heart muscle by electron spin resonance spectroscopy. Cells that overexpress CYP2E1 can be protected from arachidonic acid toxicity damage by TEMPOL.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT RE 2 Oral

Target Organs

Liver,spleen

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

294.8 °F - DIN 51758

Flash Point(C)

146 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clean and selective oxidation of alcohols catalyzed by ion-supported TEMPO in water.
Qian W, et al.
Tetrahedron, 62(4), 556-562 (2006)
Synthesis of polymer-supported TEMPO catalysts and their application in the oxidation of various alcohols.
Tanyeli C and Gumus A.
Tetrahedron Letters, 44(8), 1639-1642 (2003)
Synlett, 2767-2767 (2006)
Journal of Polymer Science. Part B, Polymer Physics, 31, 1885-1885 (1993)
T Tanigawa et al.
Free radical research communications, 19 Suppl 1, S171-S176 (1993-01-01)
A method was designed to detect free radicals in circulating blood of animals using extracorporeal circulation and electron paramagnetic resonance (EPR). An arterial-venous shunt was inserted into male Wistar rats between the distal abdominal aorta and the inferior vena cava.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service