Skip to Content
Merck
All Photos(3)

Key Documents

408220

Sigma-Aldrich

Sodium acrylate

97%

Synonym(s):

Acrylic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2Na
CAS Number:
Molecular Weight:
94.04
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

mp

>300 °C (lit.)

SMILES string

[Na+].[O-]C(=O)C=C

InChI

1S/C3H4O2.Na/c1-2-3(4)5;/h2H,1H2,(H,4,5);/q;+1/p-1

InChI key

NNMHYFLPFNGQFZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium acrylate (SA) is a metal salt that can be prepared by an acid-base reaction between sodium hydroxide and acrylic acid. It can be used in the preparation of poly(sodium acrylate) using bulk, solution, emulsion, and suspension polymerization techniques. It results in the formation of a water soluble polymer that can be used in a variety of industries and personal care applications.

Application

SA can be used in tuning the grain size of iron oxide microparticles for potential applications in biological assaying and in chemical sensors. It can be used as a reducing and capping agent on gold nanoparticles which can be used in the biological application.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymerization of sodium acrylate in inverse-suspension stabilized by sorbitan fatty esters
Omidian H, et al.
European Polymer Journal, 39(5), 1013-1018 (2003)
Preparation of acrylate-stabilized gold and silver hydrosols and gold- polymer composite films
Hussain I, et al.
Langmuir, 19(11), 4831-4835 (2003)
Aggregative growth in the size-controlled growth of monodispersed gold nanoparticles
Njoki PN, et al.
Langmuir, 26(16), 13622-13629 (2010)
Tuning the grain size and particle size of superparamagnetic Fe3O4 microparticles
Xuan S, et al.
Chemistry of Materials, 21(21), 5079-5087 (2009)
Gerlinde Lefever et al.
Materials (Basel, Switzerland), 13(2) (2020-01-18)
Addition of superabsorbent polymers (SAPs) to cementitious mixtures promotes the self-healing ability of the material. When cracking occurs; SAPs present inside the crack will swell upon contact with water and subsequently release this water to stimulate the further hydration of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service