Skip to Content
Merck
All Photos(1)

Key Documents

139564

Sigma-Aldrich

1,2,7,8-Diepoxyoctane

97%

Synonym(s):

1,7-Octadiene bisoxide, 1,7-Octadiene diepoxide, 2,2′-(1,4-Butanediyl)bis[oxirane]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O2
CAS Number:
Molecular Weight:
142.20
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.445 (lit.)

bp

240 °C (lit.)

density

0.997 g/mL at 25 °C (lit.)

SMILES string

C(CCC1CO1)CC2CO2

InChI

1S/C8H14O2/c1(3-7-5-9-7)2-4-8-6-10-8/h7-8H,1-6H2

InChI key

LFKLPJRVSHJZPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Removal of GenX by APTES functionalized diepoxyoctane cross-linked chitosan beads: Cross-linked and aminated chitosan beads were synthesized using 1,2:7,8-diepoxyoctane and 3-aminopropyl triethoxysilane. These beads were studied for their efficiency in removing GenX, a problematic environmental pollutant, demonstrating a potential application in water treatment technologies (M Vakili, F Gholami, HM Zwain, W Wang… - Journal of …, 2023 - Elsevier).

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Zhao
Journal of biomaterials science. Polymer edition, 17(4), 419-433 (2006-06-14)
Hyaluronic acid (hyaluronan, HA) has many medical applications as a biomaterial. To enhance its biostability, a novel hydrogel of cross-linked hyaluronic acid was prepared using a double cross-linking process, which involves building cross-linkages between hydroxyl group pairs and carboxyl group
E H Vock et al.
Mutation research, 441(1), 85-93 (1999-05-04)
The time-dependent dose-response relationships for the induction of DNA double-strand breaks (DSB) assessed by pulsed-field gel electrophoresis (PFGE) and for viability (evaluated by the MTT cytotoxicity test) were investigated in order to discriminate between genotoxic and cytotoxic mechanisms of DNA
A H al-Hakim et al.
The Biochemical journal, 251(3), 935-938 (1988-05-01)
A new method for the chemical labelling of nucleic acid with biotin to produce non-radioactive probes has been developed. NN'-Bis-(3-aminopropyl)butane-1,4-diamine (spermine) and long-chain diamino compounds (diaminohexane, diaminodecane and diaminododecane) were linked covalently to biotin and the resultant conjugates were attached
M J Yunes et al.
Chemical research in toxicology, 9(6), 994-1000 (1996-09-01)
The carcinogenicity of epoxide compounds has been attributed to covalent binding to DNA. Whereas monoepoxides form only monoadducts, diepoxides can form both monoadducts and interstrand cross-links. The latter are believed to be the more significant cytotoxic lesions as diepoxides are
J T Millard et al.
Biochemistry, 40(35), 10677-10685 (2001-08-29)
Diepoxyalkanes form interstrand cross-links in DNA oligomers preferentially at 5'-GNC sites. We have examined cross-linking by 1,2,3,4-diepoxybutane (DEB) and 1,2,7,8-diepoxyoctane (DEO) within a fragment of the 5S RNA gene of Xenopus borealis in both the free and nucleosomal states. Sites

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service