Skip to Content
Merck
All Photos(1)

Documents

338036

Sigma-Aldrich

Neopentyl glycol diglycidyl ether

technical grade

Synonym(s):

1,3-Bis(2,3-epoxypropoxy)-2,2-dimethylpropane, 2,2′-[(2,2-Dimethyl-1,3-propanediyl)bis(oxymethylene)]bis[oxirane]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20O4
CAS Number:
Molecular Weight:
216.27
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

vapor density

>1 (vs air)

form

liquid

eq. wt.

135-165 g (epoxide)

refractive index

n20/D 1.457 (lit.)

viscosity

10-30 mPa.s(25 °C)

bp

103-107 °C/1 mmHg (lit.)

density

1.04 g/mL at 25 °C (lit.)

SMILES string

CC(C)(COCC1CO1)COCC2CO2

InChI

1S/C11H20O4/c1-11(2,7-12-3-9-5-14-9)8-13-4-10-6-15-10/h9-10H,3-8H2,1-2H3

InChI key

KUAUJXBLDYVELT-UHFFFAOYSA-N

General description

Neopentyl glycol diglycidyl ether (NGDE) is a low-toxic epoxy molecule with two epoxy pendant groups at the end of the long chain. It has excellent reactivity, low viscosity, and cross-linking properties. It facilitates the resin with high strength and impact resistance. It is used as a precursor to synthesize hydrogels for drug delivery, dental materials, coating, and adhesives for medical devices and tissue engineering. It is also used as a solid matrix to fabricate light-emitting devices like OLEDs.

Application

Neopentyl glycol diglycidyl ether can be used,



  • As a cross-linking agent to prepare Pullulan/polydopamine hydrogels for selective adsorption of pollutant cationic dyes.
  • As a monomer to synthesize gel polymer electrolytes for Li-ion batteries via cationic ring-open polymerization.
  • As a precursor for the synthesis of hybrid epoxy methacrylate resins cationic and free-radical photopolymerization. These epoxy resins can be used as anti-corrosion and decorative coatings.
  • To synthesize epoxy nanocomposites for fabrication of light-emitting devices.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile tailoring of thermal transition temperatures of epoxy shape memory polymers
Xie T and Rousseau IA
Polymer, 50(8), 1852-1856 (2009)
Photopolymerization of neopentyl glycol diglycidylether and its compositions with 9-(2-oxiranylmethyl) carbazole
Simokaitiene J, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 147(1), 55-61 (2002)
Incorporation of dumbbell-shaped and Y-shaped cross-linkers in adjustable pullulan/polydopamine hydrogels for selective adsorption of cationic dyes
Ting Su, et al.
Environmental Research, 182, 109010-109010 (2020)
A new flexible soy-based adhesive enhanced with neopentyl glycol diglycidyl ether: Properties and application
Luo J, et al.
Polymers, 8(9), 346-346 (2016)
Investigation of the reactivity of epoxide monomers in photoinitiated cationic polymerization
Bulut U and Crivello JV
Macromolecules, 38(9), 3584-3595 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service